Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC13D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199242.2 | NP_954712.1 | 25 | 2789 | ||
XM_011524504.1 | XP_011522806.1 | 25 | 2422 | |||
XM_011524506.1 | XP_011522808.1 | 25 | 2419 | |||
XM_011524507.2 | XP_011522809.1 | 25 | 2819 | |||
GenBank mRNA | AJ578444.1 | - | 25 | 2467 | ||
AK024474.1 | - | 23 | 2691 | |||
AK300316.1 | - | 26 | 2480 | |||
AK301529.1 | - | 25 | 2422 | |||
BC024740.2 | - | 23 | 2218 | |||
BC067084.1 | - | 25 | 2789 | 5951944 |