Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRIMPOL SIRNA Assays ›
          Gene Symbol:
          PRIMPOL
          Gene Name:
          primase and DNA directed polymerase
          Gene Aliases:
          CCDC111, MYP22
          Chromosome Location:
          Chr.4: 184649613 - 184694959 on Build GRCh38
          UniGene ID:
          Hs.481307
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s47417

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRIMPOL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001300768.1 NP_001287697.1 3 508
          NM_152683.3 NP_689896.1 3 508
          XM_011531719.1 XP_011530021.1 3 385
          XM_011531720.1 XP_011530022.1 3 423
          XM_011531721.2 XP_011530023.1 3 385
          XM_011531724.2 XP_011530026.1 3 445
          XM_011531725.1 XP_011530027.1 3 359
          XM_011531729.1 XP_011530031.1 3 344
          XM_011531730.2 XP_011530032.1 3 359
          XM_017007864.1 XP_016863353.1 3 423
          XM_017007865.1 XP_016863354.1 3 423
          XM_017007866.1 XP_016863355.1 3 423
          XM_017007867.1 XP_016863356.1 3 385
          XM_017007868.1 XP_016863357.1 3 385
          XM_017007869.1 XP_016863358.1 3 385
          XM_017007870.1 XP_016863359.1 3 385
          XM_017007871.1 XP_016863360.1 3 444
          XM_017007873.1 XP_016863362.1 3 385
          XM_017007875.1 XP_016863364.1 3 344
          GenBank mRNA AK057729.1 - 3 291
          AK307317.1 - 3 344
          BC064600.1 - 3 349 5786393
          BX647575.1 - 3 396

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial DNA replication
          DNA replication, synthesis of RNA primer
          response to UV
          translesion synthesis
          replication fork processing
          chromatin binding
          DNA-directed DNA polymerase activity
          DNA primase activity
          protein binding
          manganese ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0