Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ERCC1 SIRNA Assays ›
          Gene Symbol:
          ERCC1
          Gene Name:
          ERCC excision repair 1, endonuclease non-catalytic subunit
          Gene Aliases:
          COFS4, RAD10, UV20
          Chromosome Location:
          Chr.19: 45407333 - 45478866 on Build GRCh38
          UniGene ID:
          Hs.435981
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s4786

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ERCC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166049.1 NP_001159521.1 3 265
          NM_001983.3 NP_001974.1 3 265
          NM_202001.2 NP_973730.1 2 307
          XM_005258634.1 XP_005258691.1 3 196
          XM_005258635.2 XP_005258692.1 3 666
          XM_005258636.4 XP_005258693.1 3 257
          XM_005258637.1 XP_005258694.1 3 196
          XM_011526610.2 XP_011524912.1 3 212
          XM_017026459.1 XP_016881948.1 3 193
          XM_017026460.1 XP_016881949.1 3 666
          XM_017026461.1 XP_016881950.1 4 295
          XM_017026462.1 XP_016881951.1 4 260
          XM_017026463.1 XP_016881952.1 3 252
          XM_017026464.1 XP_016881953.1 3 253
          XM_017026465.1 XP_016881954.1 3 666
          XM_017026466.1 XP_016881955.1 4 300
          GenBank mRNA AB069681.1 - 3 185
          AF001925.1 - 3 261
          AK314884.1 - 3 197
          BC008930.2 - 3 181 2824122
          BC052813.1 - 2 250 6526165
          BM789972.1 - 2 289
          BQ671113.1 - 3 265
          BT019806.1 - 2 119
          M13194.1 - 3 261
          M28650.1 - 3 261
          S94539.1 - 1 194

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          endodeoxyribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          pyrimidine dimer repair by nucleotide-excision repair
          replicative cell aging
          DNA repair
          transcription-coupled nucleotide-excision repair
          nucleotide-excision repair
          nucleotide-excision repair, preincision complex stabilization
          nucleotide-excision repair, DNA incision, 3'-to lesion
          nucleotide-excision repair, DNA incision, 5'-to lesion
          double-strand break repair
          DNA recombination
          mitotic recombination
          syncytium formation
          response to oxidative stress
          spermatogenesis
          response to nutrient
          cell proliferation
          male gonad development
          UV protection
          response to sucrose
          response to X-ray
          multicellular organism aging
          negative regulation of telomere maintenance
          nucleotide-excision repair, DNA incision
          post-embryonic hemopoiesis
          multicellular organism growth
          interstrand cross-link repair
          isotype switching
          oogenesis
          embryonic organ development
          global genome nucleotide-excision repair
          t-circle formation
          positive regulation of t-circle formation
          single-stranded DNA endodeoxyribonuclease activity
          TFIID-class transcription factor binding
          damaged DNA binding
          single-stranded DNA binding
          protein binding
          protein C-terminus binding
          protein domain specific binding
          structure-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0