Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ERG SIRNA Assays ›
          Gene Symbol:
          ERG
          Gene Name:
          ERG, ETS transcription factor
          Gene Aliases:
          erg-3, p55
          Chromosome Location:
          Chr.21: 38367261 - 38662045 on Build GRCh38
          UniGene ID:
          Hs.473819
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s4813

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ERG
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136154.1 NP_001129626.1 5, 6 689
          NM_001136155.1 NP_001129627.1 2, 3 302
          NM_001243428.1 NP_001230357.1 5, 6 712
          NM_001243429.1 NP_001230358.1 3, 4 348
          NM_001243432.2 NP_001230361.1 5, 6 689
          NM_001291391.1 NP_001278320.1 5, 6 689
          NM_004449.4 NP_004440.1 5, 6 689
          NM_182918.3 NP_891548.1 3, 4 520
          XM_011529486.2 XP_011527788.1 5, 6 952
          XM_011529487.2 XP_011527789.1 5, 6 912
          XM_017028287.1 XP_016883776.1 5, 6 954
          XM_017028288.1 XP_016883777.1 3, 4 474
          XM_017028289.1 XP_016883778.1 3, 4 474
          GenBank mRNA AK297807.1 - 5, 6 689
          AK300395.1 - 3, 4 510
          AK301277.1 - 1, 2 46
          AK303518.1 - 2, 3 275
          AK309469.1 - 5, 6 712
          AY204741.1 - 5, 6 603
          AY204742.1 - 5, 6 603
          BC040168.1 - 3, 4 499 6052140
          DA850637.1 - 3, 4 499
          DC394344.1 - 3, 4 520
          KP100545.1 - 3, 4 396

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          endocardial cushion development
          endocardial cushion to mesenchymal transition involved in heart valve formation
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          protein phosphorylation
          signal transduction
          multicellular organism development
          cell proliferation
          cell migration
          cell differentiation
          positive regulation of blood vessel remodeling
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          DNA binding
          chromatin binding
          signal transducer activity
          protein binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0