Search
Search

MKX| Gene Symbol: MKX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242702.1 | NP_001229631.1 | 7 | 1123 | ||
| NM_173576.2 | NP_775847.2 | 7 | 1126 | |||
| XM_017016105.1 | XP_016871594.1 | 7 | 1163 | |||
| XM_017016106.1 | XP_016871595.1 | 7 | 1354 | |||
| XM_017016108.1 | XP_016871597.1 | 6 | 1026 | |||
| GenBank mRNA | AK125339.1 | - | 7 | 1128 | ||
| BC036207.1 | - | 7 | 1092 | 5262515 | ||