Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DPY19L2 SIRNA Assays ›
Gene Symbol:
DPY19L2
Gene Name:
dpy-19 like 2
Gene Aliases:
SPATA34, SPGF9
Chromosome Location:
Chr.12: 63558913 - 63669201 on Build GRCh38
UniGene ID:
Hs.533644
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s49227
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DPY19L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_173812.4
NP_776173.3
15, 16
1722
XM_006719352.2
XP_006719415.1
15, 16
1648
XM_011538215.2
XP_011536517.1
13, 14
1188
XM_011538218.2
XP_011536520.1
8, 9
943
XM_017019188.1
XP_016874677.1
15, 16
1769
XM_017019189.1
XP_016874678.1
14, 15
1709
XM_017019190.1
XP_016874679.1
15, 16
1769
XM_017019191.1
XP_016874680.1
13, 14
1619
XM_017019192.1
XP_016874681.1
13, 14
1607
XM_017019193.1
XP_016874682.1
14, 15
1304
XM_017019194.1
XP_016874683.1
14, 15
1331
XM_017019195.1
XP_016874684.1
15, 16
1379
XM_017019196.1
XP_016874685.1
15, 16
1769
XM_017019197.1
XP_016874686.1
14, 15
1602
XM_017019198.1
XP_016874687.1
15, 16
1769
XM_017019199.1
XP_016874688.1
11, 12
1392
XM_017019201.1
XP_016874690.1
14, 15
1565
XM_017019202.1
XP_016874691.1
13, 14
1178
XM_017019203.1
XP_016874692.1
13, 14
1400
XM_017019204.1
XP_016874693.1
14, 15
1638
XM_017019205.1
XP_016874694.1
11, 12
1492
GenBank mRNA
AK057511.1
-
15, 16
1693
AK310102.1
-
13, 14
1512
AL833344.1
-
15, 16
1649
AY358792.1
-
15, 16
1643
BC031225.1
-
9, 10
724
BC068442.1
-
15, 16
1725
5297524
BC125215.1
-
15, 16
1612
BC125216.1
-
15, 16
1612
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
spermatid development
protein C-linked glycosylation via 2'-alpha-mannosyl-L-tryptophan
mannosyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0