Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACSL1 SIRNA Assays ›
          Gene Symbol:
          ACSL1
          Gene Name:
          acyl-CoA synthetase long-chain family member 1
          Gene Aliases:
          ACS1, FACL1, FACL2, LACS, LACS1, LACS2
          Chromosome Location:
          Chr.4: 184755595 - 184826593 on Build GRCh38
          UniGene ID:
          Hs.406678
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s4996
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACSL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001286708.1 NP_001273637.1 7 799
          NM_001286710.1 NP_001273639.1 7 961
          NM_001286711.1 NP_001273640.1 6 796
          NM_001286712.1 NP_001273641.1 8 1144
          NM_001995.3 NP_001986.2 7 898
          XM_005262828.1 XP_005262885.1 7 726
          XM_005262829.1 XP_005262886.1 7 793
          XM_005262831.1 XP_005262888.1 7 910
          XM_011531742.1 XP_011530044.1 7 826
          XM_017007887.1 XP_016863376.1 7 768
          XM_017007888.1 XP_016863377.1 7 852
          XM_017007889.1 XP_016863378.1 7 910
          GenBank mRNA AK292798.1 - 7 752
          AK296550.1 - 8 994
          AK296826.1 - 7 961
          AK303486.1 - 6 716
          BC026290.1 - 2 192
          BC050073.1 - 7 799 5267238
          D10040.1 - 6 680
          DQ083029.1 - 6 701
          L09229.1 - 7 740

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00242530_m1

          Panther Classification:

          Molecular Function -

          ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          long-chain fatty acid metabolic process
          triglyceride metabolic process
          response to nutrient
          lipid biosynthetic process
          response to organic cyclic compound
          fatty acid transport
          adiponectin-activated signaling pathway
          response to oleic acid
          long-chain fatty-acyl-CoA biosynthetic process
          alpha-linolenic acid metabolic process
          xenobiotic catabolic process
          response to drug
          linoleic acid metabolic process
          long-chain fatty acid import
          positive regulation of protein serine/threonine kinase activity
          long-chain fatty acid-CoA ligase activity
          ATP binding
          decanoate--CoA ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0