Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BLM SIRNA Assays ›
          Gene Symbol:
          BLM
          Gene Name:
          Bloom syndrome RecQ like helicase
          Gene Aliases:
          BS, RECQ2, RECQL2, RECQL3
          Chromosome Location:
          Chr.15: 90717327 - 90815462 on Build GRCh38
          UniGene ID:
          Hs.725208
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s501285

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BLM
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000057.3 NP_000048.1 17 3347
          NM_001287246.1 NP_001274175.1 18 3457
          NM_001287247.1 NP_001274176.1 17 3347
          NM_001287248.1 NP_001274177.1 17 3513
          XM_006720632.2 XP_006720695.1 11 1406
          XM_011521881.2 XP_011520183.1 14 2382
          GenBank mRNA AK295194.1 - 16 2413
          AK303159.1 - 11 1258
          AK314262.1 - 17 3276
          BC093622.1 - 17 3270 7939467
          BC101567.1 - 17 3270 8069073
          BC115030.1 - 17 3295 7961413
          BC115032.1 - 17 3295 7961425
          BC143280.1 - 17 3435 9051783
          U39817.1 - 17 3303

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA helicase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cyclin-dependent protein serine/threonine kinase activity
          double-strand break repair via homologous recombination
          DNA double-strand break processing
          DNA synthesis involved in DNA repair
          strand displacement
          DNA strand renaturation
          DNA replication
          DNA repair
          DNA recombination
          cellular response to DNA damage stimulus
          mitotic G2 DNA damage checkpoint
          response to X-ray
          protein sumoylation
          replication fork processing
          DNA duplex unwinding
          positive regulation of transcription, DNA-templated
          negative regulation of DNA recombination
          replication fork protection
          protein oligomerization
          negative regulation of cell division
          cellular response to ionizing radiation
          cellular response to hydroxyurea
          cellular response to camptothecin
          regulation of signal transduction by p53 class mediator
          bubble DNA binding
          p53 binding
          single-stranded DNA binding
          ATP-dependent DNA helicase activity
          helicase activity
          protein binding
          ATP binding
          ATP-dependent helicase activity
          four-way junction helicase activity
          ATPase activity
          annealing helicase activity
          ATP-dependent 3'-5' DNA helicase activity
          G-quadruplex DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0