Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FGFR3 SIRNA Assays ›
          Gene Symbol:
          FGFR3
          Gene Name:
          fibroblast growth factor receptor 3
          Gene Aliases:
          ACH, CD333, CEK2, HSFGFR3EX, JTK4
          Chromosome Location:
          Chr.4: 1793299 - 1808872 on Build GRCh38
          UniGene ID:
          Hs.1420
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s501308

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FGFR3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000142.4 NP_000133.1 10 1547
          NM_001163213.1 NP_001156685.1 10 1553
          NM_022965.3 NP_075254.1 8 1211
          XM_006713868.1 XP_006713931.1 10 1572
          XM_006713869.1 XP_006713932.1 10 1572
          XM_006713870.1 XP_006713933.1 10 1569
          XM_006713871.1 XP_006713934.1 10 1566
          XM_006713872.1 XP_006713935.1 10 1563
          XM_006713873.1 XP_006713936.1 10 1560
          XM_011513420.1 XP_011511722.1 10 1566
          XM_011513422.1 XP_011511724.1 10 1563
          GenBank mRNA AB209441.1 - 10 1555
          AF245114.1 - 7 994
          BC121175.2 - 3 314
          BC128610.1 - 3 314
          BC153824.1 - 3 314
          HM244683.1 - 10 1442
          KJ534834.1 - 9 1294
          M58051.1 - 9 1330
          M64347.1 - 8 1066

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          skeletal system development
          endochondral ossification
          chondrocyte differentiation
          endochondral bone growth
          JAK-STAT cascade
          cell-cell signaling
          positive regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          positive regulation of phospholipase activity
          regulation of phosphatidylinositol 3-kinase signaling
          peptidyl-tyrosine phosphorylation
          bone mineralization
          chondrocyte proliferation
          phosphatidylinositol-3-phosphate biosynthetic process
          positive regulation of tyrosine phosphorylation of Stat1 protein
          positive regulation of tyrosine phosphorylation of Stat3 protein
          positive regulation of MAPK cascade
          positive regulation of GTPase activity
          positive regulation of phosphatidylinositol 3-kinase activity
          protein autophosphorylation
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          negative regulation of developmental growth
          bone morphogenesis
          positive regulation of ERK1 and ERK2 cascade
          bone maturation
          fibroblast growth factor receptor apoptotic signaling pathway
          protein tyrosine kinase activity
          fibroblast growth factor-activated receptor activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          ATP binding
          1-phosphatidylinositol-3-kinase activity
          fibroblast growth factor binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0