Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FKTN SIRNA Assays ›
Gene Symbol:
FKTN
Gene Name:
fukutin
Gene Aliases:
CMD1X, FCMD, LGMD2M, MDDGA4, MDDGB4, MDDGC4
Chromosome Location:
Chr.9: 105558117 - 105655950 on Build GRCh38
UniGene ID:
Hs.55777
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s502523
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FKTN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079802.1
NP_001073270.1
7
961
NM_001198963.1
NP_001185892.1
7
961
NM_006731.2
NP_006722.2
6
869
XM_006717014.2
XP_006717077.1
6
873
XM_011518368.2
XP_011516670.1
8
1109
XM_011518369.2
XP_011516671.1
7
959
XM_011518370.2
XP_011516672.1
8
2645
XM_011518371.2
XP_011516673.1
8
1144
XM_011518373.2
XP_011516675.1
8
1055
XM_011518374.2
XP_011516676.1
6
873
XM_011518375.2
XP_011516677.1
6
1188
XM_011518376.2
XP_011516678.1
6
1011
XM_011518378.2
XP_011516680.1
8
1109
XM_011518379.2
XP_011516681.1
6
755
XM_011518381.2
XP_011516683.1
7
944
XM_011518387.2
XP_011516689.1
8
1109
XM_011518389.2
XP_011516691.1
8
1109
XM_011518390.2
XP_011516692.1
4
550
XM_011518391.2
XP_011516693.1
8
1109
XM_017014462.1
XP_016869951.1
9
1243
XM_017014463.1
XP_016869952.1
6
1332
XM_017014464.1
XP_016869953.1
8
1109
XM_017014465.1
XP_016869954.1
7
959
XM_017014466.1
XP_016869955.1
8
1109
XM_017014467.1
XP_016869956.1
7
1368
XM_017014468.1
XP_016869957.1
6
1251
XM_017014469.1
XP_016869958.1
8
1109
XM_017014470.1
XP_016869959.1
8
1109
XM_017014471.1
XP_016869960.1
9
1207
XM_017014472.1
XP_016869961.1
9
1233
XM_017014473.1
XP_016869962.1
4
3111
XM_017014474.1
XP_016869963.1
6
872
XM_017014475.1
XP_016869964.1
6
876
XM_017014476.1
XP_016869965.1
9
1227
XM_017014477.1
XP_016869966.1
8
1092
XM_017014478.1
XP_016869967.1
10
1294
XM_017014479.1
XP_016869968.1
7
991
GenBank mRNA
AB008226.1
-
6
856
AB609007.1
-
856
AK300840.1
-
7
959
AK304460.1
-
6
867
BC101808.1
-
5
771
8069314
BC112038.1
-
5
771
8327506
BC117699.1
-
5
777
40073633
BC117700.1
-
7
1019
40073634
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nervous system development
muscle organ development
negative regulation of cell proliferation
protein O-linked mannosylation
negative regulation of JNK cascade
regulation of protein glycosylation
transferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-7mgp5:80/100.66.78.208:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0