Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WASH1 SIRNA Assays ›
          Gene Symbol:
          WASH1 LOC100288778 WASH2P WASH3P WASH7P
          Gene Name:
          WAS protein family homolog 1
          WAS protein family homolog 1 pseudogene
          WAS protein family homolog 2 pseudogene
          WAS protein family homolog 3 pseudogene
          WAS protein family homolog 7 pseudogene
          Gene Aliases:
          FAM39E, WASH
          -
          FAM39B
          FAM39DP
          FAM39F, WASH5P
          Chromosome Location:
          Chr.9: 14475 - 30487 on Build GRCh38
          Chromosome Location:
          Chr.12: 14477 - 32015 on Build GRCh38
          Chromosome Location:
          Chr.2: 113583653 - 113599037 on Build GRCh38
          Chromosome Location:
          Chr.15: 101960813 - 101976605 on Build GRCh38
          Chromosome Location:
          Chr.1: 14362 - 29370 on Build GRCh38
          UniGene ID:
          Hs.446466
          Hs.459573
          Hs.446466
          Hs.459573
          Hs.446466
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID s51764

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WASH1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_182905.4 NP_878908.4 5 610
          XM_011517658.2 XP_011515960.1 5 727
          XM_011517659.2 XP_011515961.1 5 509
          XM_011517660.2 XP_011515962.1 5 506
          XM_011517662.2 XP_011515964.1 5 2013
          XM_011517663.2 XP_011515965.1 4 1862
          XM_011517664.2 XP_011515966.1 4 1859
          XM_011517665.2 XP_011515967.1 4 408
          XM_011517666.2 XP_011515968.1 4 407
          XM_011517671.2 XP_011515973.2 5 727
          XM_017014169.1 XP_016869658.1 5 626
          XM_017014170.1 XP_016869659.1 5 689
          XM_017014171.1 XP_016869660.1 4 437
          XM_017014172.1 XP_016869661.1 4 2015
          XM_017014173.1 XP_016869662.1 5 727
          GenBank mRNA AK024481.1 - 1 1050
          AK289708.1 - 5 613
          AK309568.1 - 4 342
          Gene Symbol: LOC100288778
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_130745.1 5 610
          GenBank mRNA CN404396.1 - 4 474
          DA015248.1 - 5 496
          Gene Symbol: WASH2P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024077.2 5 735
          GenBank mRNA AK292759.1 - 4 338
          AK308574.1 - 4 344
          AL137714.1 - 5 622
          AY217347.1 - 4 1167
          BC082240.1 - 3 1192
          BC106020.1 - 2 159
          BX377099.2 - 5 498
          BX537637.1 - 5 736
          Gene Symbol: WASH3P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003659.2 5 1284
          GenBank mRNA AK056232.1 - 5 1129
          AK057951.1 - 7 734
          AK095183.1 - 5 491
          AK291582.1 - 5 506
          AK295256.1 - 5 503
          AK300161.1 - 6 706
          BC048328.1 - 4 484
          BC063555.1 - 5 686
          BC110996.1 - 5 986
          BQ722707.1 - 4 494
          Gene Symbol: WASH7P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024540.1 5 509
          GenBank mRNA AK092583.1 - 4 384
          AK294377.1 - 4 405
          AK296182.1 - 6 645
          AK302089.1 - 5 563
          AK308540.1 - 7 764
          AK309926.1 - 5 496
          AK310121.1 - 5 383
          AK310139.1 - 6 537
          AK316487.1 - 4 698
          BC063470.1 - 3 558
          BC094698.1 - 3 1047
          DA054875.1 - 5 517
          DA143319.1 - 5 522

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          protein transport
          endosomal transport
          extracellular matrix disassembly
          positive regulation of cell migration
          positive regulation of pseudopodium assembly
          Arp2/3 complex-mediated actin nucleation
          retrograde transport, endosome to Golgi
          retrograde transport, endosome to plasma membrane
          protein targeting to lysosome
          endosome organization
          early endosome to late endosome transport
          actin binding
          protein binding
          ubiquitin protein ligase binding
          alpha-tubulin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0