Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WASH1 SIRNA Assays ›
          Gene Symbol:
          WASH1 LOC100288778 WASH2P WASH3P WASH7P
          Gene Name:
          WAS protein family homolog 1
          WAS protein family homolog 1 pseudogene
          WAS protein family homolog 2 pseudogene
          WAS protein family homolog 3 pseudogene
          WAS protein family homolog 7 pseudogene
          Gene Aliases:
          FAM39E, WASH
          -
          FAM39B
          FAM39DP
          FAM39F, WASH5P
          Chromosome Location:
          Chr.9: 14475 - 30487 on Build GRCh38
          Chromosome Location:
          Chr.12: 14477 - 32015 on Build GRCh38
          Chromosome Location:
          Chr.2: 113583653 - 113599037 on Build GRCh38
          Chromosome Location:
          Chr.15: 101960813 - 101976605 on Build GRCh38
          Chromosome Location:
          Chr.1: 14362 - 29370 on Build GRCh38
          UniGene ID:
          Hs.446466
          Hs.459573
          Hs.446466
          Hs.459573
          Hs.446466
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID s51765

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WASH1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_182905.4 NP_878908.4 5, 6 681
          XM_011517658.2 XP_011515960.1 5, 6 798
          XM_011517659.2 XP_011515961.1 5, 6 580
          XM_011517660.2 XP_011515962.1 5, 6 577
          XM_011517662.2 XP_011515964.1 5, 6 2084
          XM_011517663.2 XP_011515965.1 4, 5 1933
          XM_011517664.2 XP_011515966.1 4, 5 1930
          XM_011517665.2 XP_011515967.1 4, 5 479
          XM_011517666.2 XP_011515968.1 4, 5 478
          XM_017014169.1 XP_016869658.1 5, 6 697
          XM_017014170.1 XP_016869659.1 5, 6 760
          GenBank mRNA AK024481.1 - 1 1121
          AK289708.1 - 5, 6 684
          AK309568.1 - 4 413
          Gene Symbol: LOC100288778
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_130745.1 5, 6 681
          GenBank mRNA BC063682.1 - 1, 2 27
          CN404396.1 - 4, 5 545
          Gene Symbol: WASH2P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK292759.1 - 4, 5 409
          AK308574.1 - 4 415
          AL137714.1 - 5 693
          AY217347.1 - 4, 5 1238
          BC106020.1 - 2, 3 230
          BX537637.1 - 5, 6 807
          Gene Symbol: WASH3P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003659.2 5, 6 1355
          GenBank mRNA AK056232.1 - 5, 6 1200
          AK057951.1 - 7, 8 805
          AK095183.1 - 5 562
          AK291582.1 - 5 577
          AK295256.1 - 5 574
          AK300161.1 - 6, 7 777
          BC048328.1 - 4, 5 555
          BC063555.1 - 5 757
          BQ722707.1 - 4, 5 565
          Gene Symbol: WASH7P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024540.1 5, 6 580
          GenBank mRNA AK092583.1 - 4, 5 455
          AK294377.1 - 4 476
          AK296182.1 - 6, 7 716
          AK302089.1 - 5, 6 634
          AK310121.1 - 5, 6 454
          AK310139.1 - 6, 7 608
          AK316487.1 - 4, 5 769
          BC094698.1 - 3 1118

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          protein transport
          endosomal transport
          extracellular matrix disassembly
          positive regulation of cell migration
          positive regulation of pseudopodium assembly
          Arp2/3 complex-mediated actin nucleation
          retrograde transport, endosome to Golgi
          retrograde transport, endosome to plasma membrane
          protein targeting to lysosome
          endosome organization
          early endosome to late endosome transport
          actin binding
          protein binding
          ubiquitin protein ligase binding
          alpha-tubulin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0