Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HES5 SIRNA Assays ›
          Gene Symbol:
          HES5
          Gene Name:
          hes family bHLH transcription factor 5
          Gene Aliases:
          bHLHb38
          Chromosome Location:
          Chr.1: 2528745 - 2530554 on Build GRCh38
          UniGene ID:
          Hs.57971
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s52198

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HES5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001010926.3 NP_001010926.1 3 1030
          XM_005244751.4 XP_005244808.1 2 1435
          GenBank mRNA BC087840.1 - 1 1225 6204648
          BX104111.1 - 1 226
          DQ272660.1 - 3 1030

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          chondrocyte differentiation
          transcription, DNA-templated
          protein complex assembly
          cell adhesion
          Notch signaling pathway
          smoothened signaling pathway
          brain development
          positive regulation of cell proliferation
          oligodendrocyte development
          telencephalon development
          glial cell fate commitment
          forebrain radial glial cell differentiation
          neural tube development
          central nervous system myelination
          positive regulation of BMP signaling pathway
          regulation of myelination
          auditory receptor cell differentiation
          positive regulation of tyrosine phosphorylation of Stat3 protein
          auditory receptor cell fate determination
          camera-type eye development
          regulation of cell differentiation
          regulation of auditory receptor cell differentiation
          negative regulation of neuron differentiation
          positive regulation of Notch signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JAK-STAT cascade
          cell maturation
          positive regulation of smooth muscle cell proliferation
          astrocyte differentiation
          negative regulation of astrocyte differentiation
          negative regulation of oligodendrocyte differentiation
          regulation of epithelial cell proliferation
          regulation of neurogenesis
          cartilage development
          inner ear receptor stereocilium organization
          comma-shaped body morphogenesis
          S-shaped body morphogenesis
          specification of loop of Henle identity
          metanephric nephron tubule morphogenesis
          establishment of epithelial cell polarity
          neuronal stem cell population maintenance
          negative regulation of stem cell differentiation
          negative regulation of pro-B cell differentiation
          negative regulation of forebrain neuron differentiation
          negative regulation of inner ear receptor cell differentiation
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          chromatin binding
          double-stranded DNA binding
          transcription factor binding
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0