Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PLOD3 SIRNA Assays ›
          Gene Symbol:
          PLOD3
          Gene Name:
          procollagen-lysine,2-oxoglutarate 5-dioxygenase 3
          Gene Aliases:
          LH3
          Chromosome Location:
          Chr.7: 101205977 - 101217730 on Build GRCh38
          UniGene ID:
          Hs.153357
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s527026

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PLOD3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001084.4 NP_001075.1 19 2653
          GenBank mRNA AF046889.1 - 19 2413
          AF068229.1 - 19 2258
          AK075338.1 - 19 2501
          AK312743.1 - 19 2411
          AL049952.1 - 9 1078
          AY220458.1 - 19 2261
          BC011674.2 - 19 2510 4300048
          BC018946.2 - 14 1575
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BM823270.1 - 3 328 Chr. 7 - 101206132 - 101208918 [-] on Build GRCh38
          GenBankmRNA BU151410.1 - 4 520 Chr. 7 - 101205984 - 101208960 [-] on Build GRCh38
          GenBankmRNA W96281.1 - 398 Chr. - - [-] on Build
          GenBankmRNA BQ720158.1 - 85 Chr. - - [-] on Build
          GenBankmRNA CN348164.1 - 3 434 Chr. 7 - 101206145 - 101207652 [-] on Build GRCh38
          GenBankmRNA AJ708820.1 - 1 173 Chr. 7 - 101205989 - 101206438 [-] on Build GRCh38
          GenBankmRNA BE002604.1 - 1 45 Chr. 7 - 101205992 - 101206330 [-] on Build GRCh38
          GenBankmRNA BM128557.1 - 4 443 Chr. 7 - 101206284 - 101208886 [-] on Build GRCh38
          GenBankmRNA CR745471.1 - 3 279 Chr. 7 - 101206076 - 101207594 [-] on Build GRCh38
          GenBankmRNA DB538169.1 - 1 118 Chr. 7 - 101205978 - 101206418 [-] on Build GRCh38
          GenBankmRNA BI905807.1 - 41 Chr. - - [-] on Build
          GenBankmRNA CX754476.1 - 5 578 Chr. 7 - 101206083 - 101210156 [-] on Build GRCh38
          GenBankmRNA BM128543.1 - 4 443 Chr. 7 - 101206218 - 101208886 [-] on Build GRCh38
          GenBankmRNA BE926594.1 - 2 209 Chr. 7 - 101206274 - 101206852 [-] on Build GRCh38
          GenBankmRNA BM555335.1 - 7 787 Chr. 7 - 101206178 - 101210621 [-] on Build GRCh38
          GenBankmRNA BM050666.1 - 378 Chr. - - [-] on Build
          GenBankmRNA BQ425227.1 - 6 633 Chr. 7 - 101206084 - 101210357 [-] on Build GRCh38
          GenBankmRNA AA340606.1 - 2 247 Chr. 7 - 101206205 - 101206889 [-] on Build GRCh38
          GenBankmRNA BM128043.1 - 4 442 Chr. 7 - 101206212 - 101208885 [-] on Build GRCh38
          GenBankmRNA BG036313.1 - 1 27 Chr. 7 - 101205984 - 101206327 [-] on Build GRCh38
          GenBankmRNA DB524073.1 - 1 146 Chr. 7 - 101205987 - 101206438 [-] on Build GRCh38
          GenBankmRNA DB557044.1 - 1 127 Chr. 7 - 101205978 - 101206427 [-] on Build GRCh38
          GenBankmRNA BM128084.1 - 4 444 Chr. 7 - 101206211 - 101208886 [-] on Build GRCh38
          GenBankmRNA BQ286408.1 - 3 297 Chr. 7 - 101206100 - 101207605 [-] on Build GRCh38
          GenBankmRNA BF968182.1 - 3 417 Chr. 7 - 101206030 - 101207710 [-] on Build GRCh38
          GenBankmRNA CA942114.1 - 2 202 Chr. 7 - 101206100 - 101206844 [-] on Build GRCh38
          GenBankmRNA BM707423.1 - 2 183 Chr. 7 - 101205989 - 101206817 [-] on Build GRCh38
          GenBankmRNA BG469256.1 - 3 394 Chr. 7 - 101106789 - 101207709 [-] on Build GRCh38
          GenBankmRNA BM792553.1 - 5 582 Chr. 7 - 101206256 - 101210146 [-] on Build GRCh38
          GenBankmRNA BU539074.1 - 307 Chr. - - [-] on Build
          GenBankmRNA BG993055.1 - 2 242 Chr. 7 - 101206255 - 101206871 [-] on Build GRCh38
          GenBankmRNA BM851151.1 - 3 278 Chr. 7 - 101206290 - 101207593 [-] on Build GRCh38
          GenBankmRNA AW385059.1 - 4 478 Chr. 7 - 101206263 - 101208922 [-] on Build GRCh38
          GenBankmRNA BP431037.1 - 3 300 Chr. 7 - 101206151 - 101207609 [-] on Build GRCh38
          GenBankmRNA AA131112.1 - 1 101 Chr. 7 - 101205988 - 101206401 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          endothelial cell morphogenesis
          protein localization
          neural tube development
          collagen fibril organization
          cellular response to hormone stimulus
          vasodilation
          epidermis morphogenesis
          oxidation-reduction process
          lung morphogenesis
          basement membrane assembly
          iron ion binding
          protein binding
          procollagen-lysine 5-dioxygenase activity
          L-ascorbic acid binding
          procollagen glucosyltransferase activity
          procollagen galactosyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0