Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PLOD3 SIRNA Assays ›
          Gene Symbol:
          PLOD3
          Gene Name:
          procollagen-lysine,2-oxoglutarate 5-dioxygenase 3
          Gene Aliases:
          LH3
          Chromosome Location:
          Chr.7: 101205977 - 101217730 on Build GRCh38
          UniGene ID:
          Hs.153357
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s527027

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PLOD3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001084.4 NP_001075.1 18 2472
          GenBank mRNA AF046889.1 - 18 2232
          AF068229.1 - 18 2077
          AK075338.1 - 18 2320
          AK312743.1 - 18 2230
          AL049952.1 - 8 897
          AY220458.1 - 18 2080
          BC011674.2 - 18 2329 4300048
          BC018946.2 - 13 1394
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BM823270.1 - 2 147 Chr. 7 - 101206132 - 101208918 [-] on Build GRCh38
          GenBankmRNA BU151410.1 - 3 339 Chr. 7 - 101205984 - 101208960 [-] on Build GRCh38
          GenBankmRNA W96281.1 - 212 Chr. - - [-] on Build
          GenBankmRNA CN348164.1 - 2 253 Chr. 7 - 101206145 - 101207652 [-] on Build GRCh38
          GenBankmRNA CR745471.1 - 2 98 Chr. 7 - 101206076 - 101207594 [-] on Build GRCh38
          GenBankmRNA CX754476.1 - 4 397 Chr. 7 - 101206083 - 101210156 [-] on Build GRCh38
          GenBankmRNA BM128557.1 - 3 262 Chr. 7 - 101206284 - 101208886 [-] on Build GRCh38
          GenBankmRNA BM128543.1 - 3 262 Chr. 7 - 101206218 - 101208886 [-] on Build GRCh38
          GenBankmRNA BM050666.1 - 197 Chr. - - [-] on Build
          GenBankmRNA BM555335.1 - 6 606 Chr. 7 - 101206178 - 101210621 [-] on Build GRCh38
          GenBankmRNA BQ425227.1 - 5 452 Chr. 7 - 101206084 - 101210357 [-] on Build GRCh38
          GenBankmRNA AA340606.1 - 1 66 Chr. 7 - 101206205 - 101206889 [-] on Build GRCh38
          GenBankmRNA BM128043.1 - 3 261 Chr. 7 - 101206212 - 101208885 [-] on Build GRCh38
          GenBankmRNA BM128084.1 - 3 263 Chr. 7 - 101206211 - 101208886 [-] on Build GRCh38
          GenBankmRNA BF968182.1 - 2 231 Chr. 7 - 101206030 - 101207710 [-] on Build GRCh38
          GenBankmRNA BQ286408.1 - 2 116 Chr. 7 - 101206100 - 101207605 [-] on Build GRCh38
          GenBankmRNA CA942114.1 - 1 21 Chr. 7 - 101206100 - 101206844 [-] on Build GRCh38
          GenBankmRNA BG469256.1 - 2 213 Chr. 7 - 101106789 - 101207709 [-] on Build GRCh38
          GenBankmRNA BM792553.1 - 4 401 Chr. 7 - 101206256 - 101210146 [-] on Build GRCh38
          GenBankmRNA BU539074.1 - 126 Chr. - - [-] on Build
          GenBankmRNA BG993055.1 - 1 57 Chr. 7 - 101206255 - 101206871 [-] on Build GRCh38
          GenBankmRNA BM851151.1 - 2 97 Chr. 7 - 101206290 - 101207593 [-] on Build GRCh38
          GenBankmRNA AW385059.1 - 3 297 Chr. 7 - 101206263 - 101208922 [-] on Build GRCh38
          GenBankmRNA BP431037.1 - 2 119 Chr. 7 - 101206151 - 101207609 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          endothelial cell morphogenesis
          protein localization
          neural tube development
          collagen fibril organization
          cellular response to hormone stimulus
          vasodilation
          epidermis morphogenesis
          oxidation-reduction process
          lung morphogenesis
          basement membrane assembly
          iron ion binding
          protein binding
          procollagen-lysine 5-dioxygenase activity
          L-ascorbic acid binding
          procollagen glucosyltransferase activity
          procollagen galactosyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0