Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APOA5 SIRNA Assays ›
          Gene Symbol:
          APOA5
          Gene Name:
          apolipoprotein A5
          Gene Aliases:
          APOAV, RAP3
          Chromosome Location:
          Chr.11: 116789370 - 116792420 on Build GRCh38
          UniGene ID:
          Hs.283923
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s528180

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: APOA5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166598.1 NP_001160070.1 4 233
          NM_052968.4 NP_443200.2 4 257
          XM_017017169.1 XP_016872658.1 3 715
          GenBank mRNA AF202889.1 - 3 192
          AF202890.1 - 3 192
          AK292739.1 - 3 204
          AK308047.1 - 2 693
          AK315229.1 - 3 204
          AY358749.1 - 3 195
          BC101787.1 - 3 220 8069293
          BC101789.1 - 3 220 8069295
          DA641693.1 - 4 233
          DB184692.1 - 3 228

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          triglyceride metabolic process
          cholesterol biosynthetic process
          lipid transport
          positive regulation of cholesterol esterification
          positive regulation of triglyceride catabolic process
          positive regulation of very-low-density lipoprotein particle remodeling
          triglyceride catabolic process
          regulation of intestinal cholesterol absorption
          neuron projection regeneration
          cholesterol efflux
          phospholipid efflux
          triglyceride-rich lipoprotein particle remodeling
          high-density lipoprotein particle assembly
          lipoprotein metabolic process
          tissue regeneration
          cholesterol homeostasis
          reverse cholesterol transport
          positive regulation of fatty acid biosynthetic process
          phosphatidylcholine metabolic process
          positive regulation of receptor-mediated endocytosis
          positive regulation of lipid catabolic process
          positive regulation of lipoprotein lipase activity
          acylglycerol homeostasis
          triglyceride homeostasis
          phospholipid binding
          enzyme activator activity
          heparin binding
          lipid binding
          cholesterol binding
          cholesterol transporter activity
          enzyme binding
          phosphatidylcholine binding
          lipase binding
          low-density lipoprotein particle receptor binding
          phosphatidylcholine-sterol O-acyltransferase activator activity
          lipase activator activity
          lipoprotein lipase activator activity
          lipoprotein particle receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0