Search
Search

EXT2| Gene Symbol: EXT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000401.3 | NP_000392.3 | 14 | 2253 | ||
| NM_001178083.1 | NP_001171554.1 | 15 | 2462 | |||
| NM_207122.1 | NP_997005.1 | 14 | 2432 | |||
| XM_011519950.1 | XP_011518252.1 | 15 | 2251 | |||
| XM_011519951.1 | XP_011518253.1 | 15 | 2361 | |||
| GenBank mRNA | AK296713.1 | - | 14 | 1893 | ||
| AK312375.1 | - | 14 | 2309 | |||
| BC010058.1 | - | 14 | 2306 | 3947503 | ||
| BC068545.1 | - | 14 | 2272 | 5263866 | ||
| BX648142.1 | - | 14 | 2230 | |||
| U62740.1 | - | 14 | 2432 | |||
| U64511.1 | - | 13 | 2124 | |||
| U72263.1 | - | 15 | 2294 | |||