Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CAPN3 SIRNA Assays ›
          Gene Symbol:
          CAPN3
          Gene Name:
          calpain 3
          Gene Aliases:
          CANP3, CANPL3, LGMD2, LGMD2A, nCL-1, p94
          Chromosome Location:
          Chr.15: 42359500 - 42412317 on Build GRCh38
          UniGene ID:
          Hs.143261
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s529147

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CAPN3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000070.2 NP_000061.1 22, 23 2695
          NM_024344.1 NP_077320.1 21, 22 2677
          NM_173087.1 NP_775110.1 19, 20 2419
          NM_173088.1 NP_775111.1 11, 12 1022
          NM_173089.1 NP_775112.1 7, 8 652
          NM_173090.1 NP_775113.1 8, 9 670
          GenBank mRNA AF127764.1 - 21, 22 2592
          AF127765.3 - 19, 20 2334
          AF147441.1 - 21
          AK027249.1 - 6, 7 1312
          BC003169.1 - 11, 12 1022
          BC003521.1 - 7, 8 634
          BC004883.2 - 7, 8 652
          BC007810.2 - 8, 9 618
          BC067126.1 - 7, 8 635
          BC100782.1 - 7, 8 633
          BC100783.1 - 7, 8 633
          BC107791.1 - 8, 9 785
          BC128605.1 - 7, 8 633
          BC146649.1 - 22, 23 2608 40147158
          BC146672.1 - 22, 23 2608 40148444
          BT007322.1 - 10, 11 853
          EU791850.1 - 22, 23 2128
          EU791851.1 - 21, 22 1984
          KU177954.1 - 10, 11 853
          KU177955.1 - 10, 11 954
          KU177956.1 - 8, 9 721
          X85030.1 - 22, 23 2389

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          protein complex assembly
          proteolysis
          apoptotic process
          signal transduction
          muscle organ development
          positive regulation of satellite cell activation involved in skeletal muscle regeneration
          response to muscle activity
          myofibril assembly
          negative regulation of protein sumoylation
          negative regulation of apoptotic process
          regulation of I-kappaB kinase/NF-kappaB signaling
          sarcomere organization
          regulation of myoblast differentiation
          positive regulation of proteolysis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          muscle cell cellular homeostasis
          regulation of catalytic activity
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of release of sequestered calcium ion into cytosol
          response to calcium ion
          muscle structure development
          G1 to G0 transition involved in cell differentiation
          cellular response to calcium ion
          cellular response to salt stress
          protein localization to membrane
          self proteolysis
          negative regulation of skeletal muscle cell differentiation
          catalytic activity
          calcium-dependent cysteine-type endopeptidase activity
          signal transducer activity
          calcium ion binding
          protein binding
          peptidase activity
          cysteine-type peptidase activity
          structural constituent of muscle
          sodium ion binding
          titin binding
          protein complex scaffold
          ligase regulator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0