Search
Search

NOS1| Gene Symbol: NOS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000620.4 | NP_000611.1 | 12 | 2671 | ||
| NM_001204213.1 | NP_001191142.1 | 11 | 1263 | |||
| NM_001204214.1 | NP_001191143.1 | 11 | 1258 | |||
| NM_001204218.1 | NP_001191147.1 | 12 | 2671 | |||
| XM_011538398.2 | XP_011536700.1 | 13 | 2542 | |||
| XM_017019345.1 | XP_016874834.1 | 12 | 2678 | |||
| XM_017019346.1 | XP_016874835.1 | 12 | 2492 | |||
| XM_017019347.1 | XP_016874836.1 | 12 | 2581 | |||
| GenBank mRNA | AK294435.1 | - | 12 | 2475 | ||
| AK307481.1 | - | 12 | 2671 | |||
| D16408.1 | - | 11 | 1960 | |||
| EU753241.1 | - | 12 | 2645 | |||
| L02881.1 | - | 11 | 2387 | |||
| U17327.1 | - | 12 | 2645 | |||
| U31466.1 | - | 11 | 1967 | |||