Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AKAP9 SIRNA Assays ›
          Gene Symbol:
          AKAP9
          Gene Name:
          A-kinase anchoring protein 9
          Gene Aliases:
          AKAP-9, AKAP350, AKAP450, CG-NAP, HYPERION, LQT11, MU-RMS-40.16A, PPP1R45, PRKA9, YOTIAO
          Chromosome Location:
          Chr.7: 91940867 - 92110673 on Build GRCh38
          UniGene ID:
          Hs.651221
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s529308

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AKAP9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005751.4 NP_005742.4 8 1646
          NM_147185.2 NP_671714.1 8 1646
          XM_017011642.1 XP_016867131.1 8 1654
          XM_017011643.1 XP_016867132.1 8 1654
          XM_017011644.1 XP_016867133.1 8 1654
          XM_017011645.1 XP_016867134.1 7 1600
          XM_017011646.1 XP_016867135.1 8 1654
          XM_017011647.1 XP_016867136.1 8 1654
          XM_017011648.1 XP_016867137.1 8 1654
          XM_017011649.1 XP_016867138.1 8 1654
          XM_017011650.1 XP_016867139.1 8 1654
          XM_017011651.1 XP_016867140.1 8 1654
          XM_017011652.1 XP_016867141.1 8 1654
          XM_017011653.1 XP_016867142.1 8 1654
          XM_017011654.1 XP_016867143.1 8 1654
          XM_017011655.1 XP_016867144.1 8 1313
          XM_017011656.1 XP_016867145.1 7 1243
          GenBank mRNA AB019691.1 - 8 1507
          AF026245.1 - 9 1641
          AF083037.1 - 1 491
          AF247727.4 - 8 1421
          AJ131693.1 - 8 1643

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          MAPK cascade
          transport
          microtubule nucleation
          signal transduction
          chemical synaptic transmission
          positive regulation of peptidyl-serine phosphorylation
          positive regulation of GTPase activity
          regulation of membrane repolarization
          regulation of ventricular cardiac muscle cell membrane repolarization
          cardiac conduction
          cellular response to cAMP
          regulation of heart rate by cardiac conduction
          positive regulation of potassium ion transmembrane transporter activity
          DNA binding
          Ras guanyl-nucleotide exchange factor activity
          receptor binding
          protein binding
          potassium channel regulator activity
          protein complex scaffold
          protein kinase A regulatory subunit binding
          ion channel binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0