Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LRRK2 SIRNA Assays ›
          Gene Symbol:
          LRRK2
          Gene Name:
          leucine rich repeat kinase 2
          Gene Aliases:
          AURA17, DARDARIN, PARK8, RIPK7, ROCO2
          Chromosome Location:
          Chr.12: 40224895 - 40369285 on Build GRCh38
          UniGene ID:
          Hs.187636
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s531025

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LRRK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_198578.3 NP_940980.3 27 3744
          XM_005268629.3 XP_005268686.1 27 3860
          XM_011537877.2 XP_011536179.1 27 3860
          XM_011537881.2 XP_011536183.1 27 3860
          XM_011537882.2 XP_011536184.1 27 3860
          XM_017018786.1 XP_016874275.1 27 3860
          XM_017018787.1 XP_016874276.1 6 688
          XM_017018789.1 XP_016874278.1 27 3860
          GenBank mRNA AK127729.1 - 18 2474
          AK131537.1 - 2 48
          AL832453.2 - 6 919
          AY792511.1 - 27 3623
          BC117180.1 - 27 3681 40125731

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPKK activity
          activation of MAPK activity
          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          protein phosphorylation
          endocytosis
          autophagy
          response to oxidative stress
          mitochondrion organization
          Golgi organization
          lysosome organization
          small GTPase mediated signal transduction
          neuromuscular junction development
          determination of adult lifespan
          cellular response to starvation
          regulation of autophagy
          positive regulation of autophagy
          regulation of protein kinase A signaling
          negative regulation of protein processing
          regulation of neuron maturation
          negative regulation of macroautophagy
          phosphorylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          calcium-mediated signaling
          olfactory bulb development
          tangential migration from the subventricular zone to the olfactory bulb
          positive regulation of protein ubiquitination
          negative regulation of protein binding
          positive regulation of protein binding
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          negative regulation of GTPase activity
          cellular response to oxidative stress
          cellular protein localization
          intracellular signal transduction
          regulation of kidney size
          exploration behavior
          locomotory exploration behavior
          regulation of lysosomal lumen pH
          regulation of locomotion
          regulation of membrane potential
          positive regulation of programmed cell death
          positive regulation of MAP kinase activity
          positive regulation of GTPase activity
          GTP metabolic process
          protein autophosphorylation
          intracellular distribution of mitochondria
          neuron projection morphogenesis
          regulation of calcium-mediated signaling
          mitochondrion localization
          regulation of mitochondrial depolarization
          regulation of synaptic transmission, glutamatergic
          canonical Wnt signaling pathway
          excitatory postsynaptic potential
          regulation of dopamine receptor signaling pathway
          positive regulation of dopamine receptor signaling pathway
          regulation of canonical Wnt signaling pathway
          regulation of dendritic spine morphogenesis
          protein localization to mitochondrion
          neuron death
          cellular response to manganese ion
          reactive oxygen species metabolic process
          regulation of mitochondrial fission
          positive regulation of canonical Wnt signaling pathway
          negative regulation of excitatory postsynaptic potential
          regulation of neuron death
          negative regulation of neuron death
          negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          positive regulation of protein autoubiquitination
          regulation of neuroblast proliferation
          regulation of synaptic vesicle transport
          negative regulation of late endosome to lysosome transport
          negative regulation of autophagosome assembly
          negative regulation of thioredoxin peroxidase activity by peptidyl-threonine phosphorylation
          negative regulation of hydrogen peroxide-induced cell death
          negative regulation of protein targeting to mitochondrion
          negative regulation of protein processing involved in protein targeting to mitochondrion
          cellular response to dopamine
          Wnt signalosome assembly
          regulation of retrograde transport, endosome to Golgi
          regulation of branching morphogenesis of a nerve
          regulation of synaptic vesicle exocytosis
          negative regulation of peroxidase activity
          SNARE binding
          glycoprotein binding
          actin binding
          GTPase activity
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase kinase activity
          GTPase activator activity
          protein binding
          ATP binding
          GTP binding
          microtubule binding
          tubulin binding
          kinase activity
          Rho GTPase binding
          syntaxin-1 binding
          receptor signaling complex scaffold activity
          clathrin binding
          GTP-dependent protein kinase activity
          peroxidase inhibitor activity
          co-receptor binding
          identical protein binding
          protein homodimerization activity
          ion channel binding
          protein kinase A binding
          beta-catenin destruction complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0