Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other POR SIRNA Assays ›
          Gene Symbol:
          POR
          Gene Name:
          cytochrome p450 oxidoreductase
          Gene Aliases:
          CPR, CYPOR, P450R
          Chromosome Location:
          Chr.7: 75915102 - 75986855 on Build GRCh38
          UniGene ID:
          Hs.354056
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s531549

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: POR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000941.2 NP_000932.3 13 1517
          GenBank mRNA AB051763.1 - 12 1426
          AB209874.1 - 12 1454
          AF258341.1 - 12 1426
          AK129978.1 - 13 1462
          AK290529.1 - 13 1497
          AK293194.1 - 7 899
          AK296096.1 - 9 1142
          AK296639.1 - 8 924
          AK304511.1 - 1752
          AK312543.1 - 13 1462
          BC034277.1 - 12 1450 3882411
          BG104767.1 - 2 53
          BX648619.1 - 13 1465
          S90469.1 - 12 1423

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of growth plate cartilage chondrocyte proliferation
          xenobiotic metabolic process
          response to nutrient
          carnitine metabolic process
          flavonoid metabolic process
          internal peptidyl-lysine acetylation
          fatty acid oxidation
          positive regulation of chondrocyte differentiation
          positive regulation of monooxygenase activity
          response to drug
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          nitrate catabolic process
          positive regulation of cholesterol biosynthetic process
          positive regulation of smoothened signaling pathway
          nitric oxide catabolic process
          oxidation-reduction process
          negative regulation of lipase activity
          demethylation
          cellular response to follicle-stimulating hormone stimulus
          cellular response to peptide hormone stimulus
          positive regulation of steroid hormone biosynthetic process
          cellular organofluorine metabolic process
          NADPH-hemoprotein reductase activity
          cytochrome-b5 reductase activity, acting on NAD(P)H
          protein binding
          nitric oxide dioxygenase activity
          electron carrier activity
          FMN binding
          hydrolase activity
          enzyme binding
          [methionine synthase] reductase activity
          iron-cytochrome-c reductase activity
          flavin adenine dinucleotide binding
          NADP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0