Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTH SIRNA Assays ›
          Gene Symbol:
          PTH
          Gene Name:
          parathyroid hormone
          Gene Aliases:
          PTH1
          Chromosome Location:
          Chr.11: 13492045 - 13496175 on Build GRCh38
          UniGene ID:
          Hs.37045
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s531725

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTH
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000315.3 NP_000306.1 3 239
          NM_001316352.1 NP_001303281.1 3 263
          GenBank mRNA BC096142.1 - 3 223 40002688
          BC096143.1 - 3 223 40002690
          BC096144.3 - 3 223 40002692
          BC096145.1 - 3 223 40002693
          BG188276.1 - 3 273
          BX103059.1 - 3 229
          BX106232.1 - 3 239
          V00597.1 - 3 198
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA W56120.1 - 213 Chr. - - [-] on Build
          GenBankmRNA W37708.1 - 239 Chr. - - [-] on Build
          GenBankmRNA W38764.1 - 203 Chr. - - [-] on Build
          GenBankmRNA BG198564.1 - 2 141 Chr. 11 - 13492062 - 13492860 [-] on Build GRCh38
          GenBankmRNA W39202.1 - 82 Chr. - - [-] on Build
          GenBankmRNA W19763.1 - 89 Chr. - - [-] on Build
          GenBankmRNA W39062.1 - 214 Chr. - - [-] on Build
          GenBankmRNA W56820.1 - 208 Chr. - - [-] on Build
          GenBankmRNA W38966.1 - 201 Chr. - - [-] on Build
          GenBankmRNA W32885.1 - 228 Chr. - - [-] on Build
          GenBankmRNA W19765.1 - 218 Chr. - - [-] on Build
          GenBankmRNA W31998.1 - 2 63 Chr. 11 - 13492279 - 13492794 [-] on Build GRCh38
          GenBankmRNA W56235.1 - 218 Chr. - - [-] on Build
          GenBankmRNA W32903.1 - 211 Chr. - - [-] on Build
          GenBankmRNA W52945.1 - 28 Chr. - - [-] on Build

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          skeletal system development
          cellular calcium ion homeostasis
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          Rho protein signal transduction
          cell-cell signaling
          hormone-mediated apoptotic signaling pathway
          positive regulation of signal transduction
          response to lead ion
          regulation of gene expression
          positive regulation of bone mineralization
          positive regulation of cAMP biosynthetic process
          response to vitamin D
          cellular macromolecule biosynthetic process
          response to drug
          bone resorption
          response to ethanol
          positive regulation of glycogen biosynthetic process
          positive regulation of transcription from RNA polymerase II promoter
          cAMP metabolic process
          positive regulation of glucose import
          response to cadmium ion
          homeostasis of number of cells within a tissue
          response to parathyroid hormone
          response to fibroblast growth factor
          positive regulation of cell proliferation in bone marrow
          negative regulation of apoptotic process in bone marrow
          positive regulation of osteoclast proliferation
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          hormone activity
          parathyroid hormone receptor binding
          type 1 parathyroid hormone receptor binding
          peptide hormone receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0