Search
Search

TGFBR2| Gene Symbol: TGFBR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024847.2 | NP_001020018.1 | 2 | 541 | ||
| XM_011534043.2 | XP_011532345.1 | 2 | 314 | |||
| GenBank mRNA | AK300383.1 | - | 2 | 442 | ||
| AK304404.1 | - | 2 | 210 | |||
| CB988859.1 | - | 2 | 469 | |||
| D28131.1 | - | 2 | 99 | |||
| KU178360.1 | - | 2 | 159 | |||