Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TGFBR2 SIRNA Assays ›
          Gene Symbol:
          TGFBR2
          Gene Name:
          transforming growth factor beta receptor 2
          Gene Aliases:
          AAT3, FAA3, LDS1B, LDS2, LDS2B, MFS2, RIIC, TAAD2, TGFR-2, TGFbeta-RII
          Chromosome Location:
          Chr.3: 30606490 - 30694142 on Build GRCh38
          UniGene ID:
          Hs.604277
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s532071

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TGFBR2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001024847.2 NP_001020018.1 5 1613
          NM_003242.5 NP_003233.4 4 1538
          XM_011534043.2 XP_011532345.1 5 1386
          XM_011534045.2 XP_011532347.1 4 1166
          XM_017007106.1 XP_016862595.1 4 2752
          GenBank mRNA AK300383.1 - 5 1514
          AK304404.1 - 6 1175
          AK314102.1 - 4 1454
          BC040499.1 - 4 1539
          BM457423.1 - 1 341
          M85079.1 - 4 1491

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          serine/threonine protein kinase receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          patterning of blood vessels
          vasculogenesis
          in utero embryonic development
          heart looping
          positive regulation of mesenchymal cell proliferation
          lens development in camera-type eye
          positive regulation of tolerance induction to self antigen
          positive regulation of B cell tolerance induction
          positive regulation of T cell tolerance induction
          outflow tract morphogenesis
          growth plate cartilage chondrocyte growth
          protein phosphorylation
          receptor-mediated endocytosis
          apoptotic process
          transforming growth factor beta receptor signaling pathway
          common-partner SMAD protein phosphorylation
          Notch signaling pathway
          smoothened signaling pathway
          gastrulation
          brain development
          heart development
          embryo implantation
          aging
          response to nutrient
          positive regulation of cell proliferation
          response to mechanical stimulus
          response to glucose
          regulation of gene expression
          positive regulation of epithelial cell migration
          positive regulation of epithelial to mesenchymal transition
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          signal transduction by protein phosphorylation
          negative regulation of transforming growth factor beta receptor signaling pathway
          organ regeneration
          activation of protein kinase activity
          embryonic hemopoiesis
          wound healing
          regulation of cell proliferation
          response to drug
          myeloid dendritic cell differentiation
          positive regulation of skeletal muscle tissue regeneration
          response to estrogen
          positive regulation of angiogenesis
          response to steroid hormone
          digestive tract development
          positive regulation of smooth muscle cell proliferation
          embryonic cranial skeleton morphogenesis
          positive regulation of NK T cell differentiation
          palate development
          negative regulation of cardiac muscle cell proliferation
          pathway-restricted SMAD protein phosphorylation
          ventricular septum morphogenesis
          bronchus morphogenesis
          trachea formation
          mammary gland morphogenesis
          lung lobe morphogenesis
          response to cholesterol
          positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
          lens fiber cell apoptotic process
          positive regulation of reactive oxygen species metabolic process
          transmembrane receptor protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          transforming growth factor beta-activated receptor activity
          transforming growth factor beta receptor activity, type II
          protein binding
          ATP binding
          glycosaminoglycan binding
          mitogen-activated protein kinase kinase kinase binding
          type I transforming growth factor beta receptor binding
          SMAD binding
          metal ion binding
          transforming growth factor beta binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0