Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GCH1 SIRNA Assays ›
          Gene Symbol:
          GCH1
          Gene Name:
          GTP cyclohydrolase 1
          Gene Aliases:
          DYT14, DYT5, DYT5a, GCH, GTP-CH-1, GTPCH1, HPABH4B
          Chromosome Location:
          Chr.14: 54842005 - 54902824 on Build GRCh38
          UniGene ID:
          Hs.86724
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s532131

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GCH1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000161.2 NP_000152.1 2, 3 618
          NM_001024024.1 NP_001019195.1 2, 3 618
          NM_001024070.1 NP_001019241.1 2, 3 618
          NM_001024071.1 NP_001019242.1 2, 3 618
          XM_017021218.1 XP_016876707.1 2, 3 1050
          GenBank mRNA AY137463.1 - 2, 3 240
          AY137464.1 - 2, 3 240
          AY137465.1 - 2, 3 240
          BC025415.2 - 2, 3 605 4129035
          CR536551.1 - 2, 3 457
          S43856.1 - 2, 3 524
          S44049.1 - 2, 3 431
          S44053.1 - 2, 3 524
          U19523.1 - 2, 3 605
          U66095.1 - 2, 3 601
          U66097.1 - 2, 3 610
          Z16418.1 - 2, 3 281
          Z29433.2 - 2, 3 556
          Z29434.2 - 2, 3 604

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          hydrolase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          tetrahydrobiopterin biosynthetic process
          nitric oxide biosynthetic process
          regulation of blood pressure
          regulation of lung blood pressure
          response to lipopolysaccharide
          response to interferon-gamma
          response to tumor necrosis factor
          7,8-dihydroneopterin 3'-triphosphate biosynthetic process
          vasodilation
          dopamine biosynthetic process
          pteridine-containing compound biosynthetic process
          negative regulation of blood pressure
          tetrahydrofolate biosynthetic process
          response to pain
          neuromuscular process controlling posture
          regulation of nitric-oxide synthase activity
          positive regulation of nitric-oxide synthase activity
          protein homooligomerization
          protein heterooligomerization
          regulation of removal of superoxide radicals
          GTP cyclohydrolase I activity
          calcium ion binding
          protein binding
          GTP binding
          zinc ion binding
          GTP-dependent protein binding
          protein homodimerization activity
          coenzyme binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0