Search
Search

ADAMTS18| Gene Symbol: ADAMTS18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001326358.1 | NP_001313287.1 | 14 | 2473 | ||
| NM_199355.3 | NP_955387.1 | 14 | 2469 | |||
| XM_011522923.1 | XP_011521225.1 | 12 | 1880 | |||
| XM_011522924.1 | XP_011521226.1 | 12 | 1876 | |||
| XM_017022988.1 | XP_016878477.1 | 11 | 1716 | |||
| XM_017022989.1 | XP_016878478.1 | 11 | 1513 | |||
| GenBank mRNA | AJ311903.1 | - | 14 | 2129 | ||
| AK131403.1 | - | 13 | 1976 | |||
| BC063283.1 | - | 14 | 2469 | 30343625 | ||