Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018122.4 | NP_060592.2 | 11, 12 | 1699 | ||
XM_006711427.3 | XP_006711490.1 | 11, 12 | 1656 | |||
XM_011509711.2 | XP_011508013.1 | 11, 12 | 1656 | |||
GenBank mRNA | AK001376.1 | - | 7, 8 | 703 | ||
AK022754.1 | - | 7, 8 | 704 | |||
AK290873.1 | - | 11, 12 | 1381 | |||
AL833633.1 | - | 11, 12 | 1384 | |||
BC045173.2 | - | 11, 12 | 1678 | 5273243 |