Search
Search

TMC8| Gene Symbol: TMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152468.4 | NP_689681.2 | 16 | 2822 | ||
| XM_011524402.2 | XP_011522704.1 | 16 | 4336 | |||
| XM_011524403.2 | XP_011522705.1 | 16 | 4323 | |||
| XM_011524404.2 | XP_011522706.1 | 16 | 4311 | |||
| XM_011524406.2 | XP_011522708.1 | 15 | 4221 | |||
| XM_017024238.1 | XP_016879727.1 | 16 | 4334 | |||
| XM_017024239.1 | XP_016879728.1 | 16 | 4265 | |||
| XM_017024240.1 | XP_016879729.1 | 16 | 4241 | |||
| XM_017024242.1 | XP_016879731.1 | 10 | 1726 | |||
| GenBank mRNA | AK090478.1 | - | 16 | 2782 | ||
| AK097987.1 | - | 1 | 553 | |||
| AK128327.1 | - | 8 | 1372 | |||