Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other STIL SIRNA Assays ›
Gene Symbol:
STIL
Gene Name:
SCL/TAL1 interrupting locus
Gene Aliases:
MCPH7, SIL
Chromosome Location:
Chr.1: 47250139 - 47314787 on Build GRCh38
UniGene ID:
Hs.525198
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s532564
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: STIL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001048166.1
NP_001041631.1
13, 14
2546
NM_001282936.1
NP_001269865.1
14, 15
2755
NM_001282937.1
NP_001269866.1
14, 15
2755
NM_001282938.1
NP_001269867.1
15, 16
2614
NM_001282939.1
NP_001269868.1
15, 16
2479
NM_003035.2
NP_003026.2
13, 14
2546
XM_006710834.3
XP_006710897.1
14, 15
2620
XM_011541991.2
XP_011540293.1
14, 15
3286
XM_011541992.2
XP_011540294.1
14, 15
3269
XM_011541994.2
XP_011540296.1
13, 14
2546
XM_011541996.2
XP_011540298.1
14, 15
2405
XM_011541998.2
XP_011540300.1
14, 15
2405
XM_011542001.2
XP_011540303.1
13, 14
2546
XM_017002123.1
XP_016857612.1
14, 15
2405
XM_017002124.1
XP_016857613.1
9, 10
1822
XM_017002125.1
XP_016857614.1
13, 14
2546
XM_017002126.1
XP_016857615.1
13, 14
2546
XM_017002127.1
XP_016857616.1
13, 14
2546
GenBank mRNA
AK128406.1
-
3, 4
2873
AK307519.1
-
13, 14
2508
BC053615.1
-
15, 16
2447
BC126223.1
-
13, 14
2622
8991939
BC144089.1
-
13, 14
2622
9052609
CR749851.1
-
13, 14
2522
M74558.1
-
14, 15
2771
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: BC108373, BC145493, BC141175, AK019471, AK049223, and others.
Gene Ontology Categories:
Function(s)
Process(es)
embryonic axis specification
in utero embryonic development
neural tube closure
heart looping
mitotic spindle organization
smoothened signaling pathway
determination of left/right symmetry
cell proliferation
neural tube development
forebrain development
notochord development
floor plate development
multicellular organism growth
negative regulation of apoptotic process
regulation of centriole replication
centrosome duplication
protein localization to centrosome
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0