Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CD59 SIRNA Assays ›
          Gene Symbol:
          CD59
          Gene Name:
          CD59 molecule
          Gene Aliases:
          16.3A5, 1F5, EJ16, EJ30, EL32, G344, HRF-20, HRF20, MAC-IP, MACIF, MEM43, MIC11, MIN1, MIN2, MIN3, MIRL, MSK21, p18-20
          Chromosome Location:
          Chr.11: 33703010 - 33736479 on Build GRCh38
          UniGene ID:
          Hs.278573
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s533126

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CD59
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000611.5 NP_000602.1 3, 4 293
          NM_001127223.1 NP_001120695.1 2, 3 459
          NM_001127225.1 NP_001120697.1 3, 4 352
          NM_001127226.1 NP_001120698.1 3, 4 287
          NM_001127227.1 NP_001120699.1 3, 4 231
          NM_203329.2 NP_976074.1 4, 5 409
          NM_203330.2 NP_976075.1 5, 6 454
          NM_203331.2 NP_976076.1 4, 5 338
          GenBank mRNA AK128571.1 - 1, 2 2250
          AK309894.1 - 2, 3 459
          AK311778.1 - 3, 4 236
          AL037045.3 - 3, 4 266
          AL703204.1 - 4, 5 356
          AU136899.1 - 4, 5 338
          AU139009.1 - 3, 4 231
          AU139541.1 - 3, 4 287
          BC001506.2 - 3, 4 217 2988140
          BI752924.1 - 5, 6 389
          BQ230563.1 - 4, 5 257
          BT007104.1 - 2, 3 177
          CR407661.1 - 2, 3 177
          CR541722.1 - 2, 3 177
          DA250543.1 - 5, 6 400
          DA578905.1 - 4, 5 353
          DA840870.1 - 3, 4 353
          DA917093.1 - 4, 5 320
          M27909.1 - 3, 4 225
          M34671.1 - 2, 3 206
          M95708.1 - 3, 4 239
          X16447.1 - 3, 4 240
          X17198.1 - 3, 4 249

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cell activation
          negative regulation of activation of membrane attack complex
          ER to Golgi vesicle-mediated transport
          cell surface receptor signaling pathway
          blood coagulation
          regulation of complement activation
          positive regulation of T cell proliferation
          negative regulation of apoptotic process
          COPII vesicle coating
          complement binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0