Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FGF14 SIRNA Assays ›
          Gene Symbol:
          FGF14
          Gene Name:
          fibroblast growth factor 14
          Gene Aliases:
          FGF-14, FHF-4, FHF4, SCA27
          Chromosome Location:
          Chr.13: 101720855 - 102402428 on Build GRCh38
          UniGene ID:
          Hs.508616
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s533673

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FGF14
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321931.1 NP_001308860.1 3, 4 573
          NM_001321932.1 NP_001308861.1 3, 4 659
          NM_001321933.1 NP_001308862.1 3, 4 569
          NM_001321934.1 NP_001308863.1 2, 3 332
          NM_001321935.1 NP_001308864.1 2, 3 185
          NM_001321936.1 NP_001308865.1 3, 4 512
          NM_001321937.1 NP_001308866.1 2, 3 678
          NM_001321938.1 NP_001308867.1 4, 5 560
          NM_001321940.1 NP_001308869.1 3, 4 915
          NM_001321941.1 NP_001308870.1 3, 4 747
          NM_001321942.1 NP_001308871.1 3, 4 754
          NM_001321943.1 NP_001308872.1 3, 4 791
          NM_001321944.1 NP_001308873.1 3, 4 1007
          NM_001321945.1 NP_001308874.1 3, 4 550
          NM_001321946.1 NP_001308875.1 4, 5 656
          NM_001321947.1 NP_001308876.1 3, 4 474
          NM_001321948.1 NP_001308877.1 3, 4 546
          NM_001321949.1 NP_001308878.1 3, 4 1243
          NM_004115.3 NP_004106.1 2, 3 309
          NM_175929.2 NP_787125.1 2, 3 420
          GenBank mRNA AY188178.1 - 2, 3 420
          BC100920.1 - 2, 3 419 40003901
          BC100921.1 - 2, 3 419 40003902
          BC100922.2 - 2, 3 419 40003903
          U66200.1 - 2, 3 309

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          signal transduction
          cell-cell signaling
          nervous system development
          regulation of synaptic plasticity
          regulation of postsynaptic membrane potential
          xenophagy
          positive regulation of high voltage-gated calcium channel activity
          regulation of synaptic vesicle recycling
          protein binding
          growth factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0