Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FOXP3 SIRNA Assays ›
          Gene Symbol:
          FOXP3
          Gene Name:
          forkhead box P3
          Gene Aliases:
          AIID, DIETER, IPEX, JM2, PIDX, XPID
          Chromosome Location:
          Chr.X: 49250436 - 49266505 on Build GRCh38
          UniGene ID:
          Hs.247700
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s533920

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FOXP3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001114377.1 NP_001107849.1 11 2179
          NM_014009.3 NP_054728.2 12 2284
          XM_006724533.2 XP_006724596.2 11 2459
          XM_011543916.2 XP_011542218.1 11 3889
          XM_017029565.1 XP_016885054.1 13 3745
          XM_017029566.1 XP_016885055.1 12 3709
          XM_017029567.1 XP_016885056.1 10 2284

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          B cell homeostasis
          cytokine production
          myeloid cell homeostasis
          CD4-positive, CD25-positive, alpha-beta regulatory T cell lineage commitment
          T cell mediated immunity
          tolerance induction to self antigen
          regulation of T cell anergy
          positive regulation of T cell anergy
          negative regulation of chronic inflammatory response
          negative regulation of T cell cytokine production
          positive regulation of peripheral T cell tolerance induction
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          negative regulation of cell proliferation
          response to virus
          negative regulation of histone deacetylation
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of interferon-gamma production
          negative regulation of interleukin-10 production
          negative regulation of interleukin-17 production
          negative regulation of interleukin-2 production
          negative regulation of interleukin-4 production
          negative regulation of interleukin-5 production
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          positive regulation of interleukin-4 production
          negative regulation of CREB transcription factor activity
          positive regulation of CD4-positive, CD25-positive, alpha-beta regulatory T cell differentiation
          positive regulation of transforming growth factor beta1 production
          positive regulation of immature T cell proliferation in thymus
          positive regulation of histone acetylation
          negative regulation of histone acetylation
          negative regulation of cytokine biosynthetic process
          T cell activation
          negative regulation of T cell proliferation
          T cell homeostasis
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of interferon-gamma biosynthetic process
          negative regulation of interleukin-2 biosynthetic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of activated T cell proliferation
          negative regulation of isotype switching to IgE isotypes
          regulation of isotype switching to IgG isotypes
          negative regulation of cytokine secretion
          negative regulation of immune response
          T cell receptor signaling pathway
          negative regulation of T-helper 17 cell differentiation
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          core promoter binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          transcription corepressor activity
          protein binding
          histone acetyltransferase binding
          protein homodimerization activity
          histone deacetylase binding
          sequence-specific DNA binding
          metal ion binding
          NF-kappaB binding
          NFAT protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0