Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LTBP4 SIRNA Assays ›
          Gene Symbol:
          LTBP4
          Gene Name:
          latent transforming growth factor beta binding protein 4
          Gene Aliases:
          ARCL1C, LTBP-4, LTBP4L, LTBP4S
          Chromosome Location:
          Chr.19: 40592888 - 40629820 on Build GRCh38
          UniGene ID:
          Hs.466766
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s534005

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LTBP4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042544.1 NP_001036009.1 28 3789
          NM_001042545.1 NP_001036010.1 25 3605
          NM_003573.2 NP_003564.2 28 3678
          XM_011527376.2 XP_011525678.1 27 3932
          XM_011527377.2 XP_011525679.1 28 3837
          XM_011527378.2 XP_011525680.1 27 3765
          XM_011527379.1 XP_011525681.1 24 3740
          XM_011527380.2 XP_011525682.1 27 3711
          XM_011527381.2 XP_011525683.1 27 3711
          XM_011527382.2 XP_011525684.1 27 3594
          XM_011527383.2 XP_011525685.1 26 3498
          XM_011527384.2 XP_011525686.1 25 3372
          XM_011527385.2 XP_011525687.1 25 3366
          XM_011527386.2 XP_011525688.1 24 3240
          XM_011527387.1 XP_011525689.1 28 3937
          XM_017027352.1 XP_016882841.1 27 3705
          XM_017027353.1 XP_016882842.1 26 3579
          XM_017027354.1 XP_016882843.1 24 3246
          GenBank mRNA AF051344.2 - 25 3605
          AF051345.2 - 28 3789
          AF054502.1 - 10 1202
          AK074499.1 - 6 1093
          AK128191.1 - 14 1787
          AK304612.1 - 3 515
          Y13622.1 - 27 3678

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          protein folding
          transmembrane receptor protein serine/threonine kinase signaling pathway
          multicellular organism development
          regulation of transforming growth factor beta receptor signaling pathway
          regulation of proteolysis
          growth hormone secretion
          regulation of cell differentiation
          transforming growth factor beta-activated receptor activity
          integrin binding
          calcium ion binding
          protein binding
          glycosaminoglycan binding
          transforming growth factor beta binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0