Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MITF SIRNA Assays ›
          Gene Symbol:
          MITF
          Gene Name:
          melanogenesis associated transcription factor
          Gene Aliases:
          CMM8, MI, WS2, WS2A, bHLHe32
          Chromosome Location:
          Chr.3: 69739435 - 69968337 on Build GRCh38
          UniGene ID:
          Hs.166017
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s534050

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MITF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000248.3 NP_000239.1 2 241
          NM_001184967.1 NP_001171896.1 3 472
          NM_001184968.1 NP_001171897.1 2 241
          NM_006722.2 NP_006713.1 3 467
          NM_198158.2 NP_937801.1 2 241
          NM_198159.2 NP_937802.1 3 602
          NM_198177.2 NP_937820.1 3 458
          XM_005264754.1 XP_005264811.1 3 574
          XM_005264755.3 XP_005264812.1 3 600
          XM_006713164.2 XP_006713227.1 4 594
          XM_011533722.2 XP_011532024.1 3 579
          XM_011533723.2 XP_011532025.1 3 599
          XM_017006444.1 XP_016861933.1 3 8789
          XM_017006445.1 XP_016861934.1 3 579
          XM_017006446.1 XP_016861935.1 3 441
          XM_017006447.1 XP_016861936.1 4 710
          XM_017006448.1 XP_016861937.1 4 594
          GenBank mRNA AB006909.1 - 3 555
          AK291318.1 - 3 574
          AK296129.1 - 3 472
          AK297858.1 - 3 404
          AL110195.1 - 3 558
          BC026961.2 - 2 171 3924101
          BC065243.1 - 2 198 6066096
          BQ219650.1 - 2 241
          DC388606.1 - 3 459
          Z29678.1 - 2 238
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DC422081.1 - 3 458 Chr. 3 - 69866224 - 69937980 [+] on Build GRCh38
          GenBankmRNA BP277367.1 - 3 460 Chr. 3 - 69866225 - 69938013 [+] on Build GRCh38
          GenBankmRNA DC361712.1 - 3 458 Chr. 3 - 69866224 - 69938033 [+] on Build GRCh38
          GenBankmRNA HY180835.1 - 3 457 Chr. 3 - 69838512 - 69937953 [+] on Build GRCh38
          GenBankmRNA DC354465.1 - 3 458 Chr. 3 - 69866224 - 69938035 [+] on Build GRCh38
          GenBankmRNA CN481686.1 - 3 425 Chr. 3 - 69866257 - 69939153 [+] on Build GRCh38
          GenBankmRNA HY163756.1 - 3 429 Chr. 3 - 69866253 - 69938011 [+] on Build GRCh38
          GenBankmRNA DC362352.1 - 3 458 Chr. 3 - 69866224 - 69938009 [+] on Build GRCh38
          GenBankmRNA DC392461.1 - 3 458 Chr. 3 - 69866224 - 69938028 [+] on Build GRCh38
          GenBankmRNA HY173803.1 - 3 454 Chr. 3 - 69838514 - 69937934 [+] on Build GRCh38
          GenBankmRNA DC362311.1 - 3 440 Chr. 3 - 69866242 - 69938042 [+] on Build GRCh38
          GenBankmRNA DC362467.1 - 3 446 Chr. 3 - 69866236 - 69938034 [+] on Build GRCh38
          GenBankmRNA BP276751.1 - 3 457 Chr. 3 - 69866224 - 69938030 [+] on Build GRCh38

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: AK052895, AK155350, BC108976, Z23066, AB221657, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          protein complex assembly
          multicellular organism development
          positive regulation of gene expression
          protein sumoylation
          melanocyte differentiation
          regulation of apoptotic process
          regulation of osteoclast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          mast cell migration
          positive regulation of DNA-templated transcription, initiation
          regulation of RNA biosynthetic process
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          protein binding
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0