Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BSCL2 SIRNA Assays ›
          Gene Symbol:
          BSCL2
          Gene Name:
          BSCL2, seipin lipid droplet biogenesis associated
          Gene Aliases:
          GNG3LG, HMN5, PELD, SPG17
          Chromosome Location:
          Chr.11: 62690262 - 62709619 on Build GRCh38
          UniGene ID:
          Hs.533709
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s534090

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BSCL2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_037948.1 1 318
          NR_037949.1 1 318
          GenBank mRNA AK315423.1 - 1 235
          BC009866.2 - 1 213
          BP370169.1 - 1 317
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA969571.1 - 1 235 Chr. 11 - 62707210 - 62709536 [-] on Build GRCh38
          GenBankmRNA BQ222204.1 - 1 201 Chr. 11 - 62705346 - 62709502 [-] on Build GRCh38
          GenBankmRNA CA941537.1 - 1 165 Chr. 11 - 62707183 - 62709466 [-] on Build GRCh38
          GenBankmRNA BX375495.2 - 1 212 Chr. 11 - 62705300 - 62709513 [-] on Build GRCh38
          GenBankmRNA DB016639.1 - 1 177 Chr. 11 - 62707106 - 62709478 [-] on Build GRCh38
          GenBankmRNA DB107899.1 - 1 237 Chr. 11 - 62707145 - 62709537 [-] on Build GRCh38
          GenBankmRNA BG715990.1 - 1 227 Chr. 11 - 62705495 - 62709526 [-] on Build GRCh38
          GenBankmRNA BQ213986.1 - 1 227 Chr. 11 - 62705348 - 62709517 [-] on Build GRCh38
          GenBankmRNA CB106765.1 - 1 218 Chr. 11 - 62707208 - 62709519 [-] on Build GRCh38
          GenBankmRNA DB055750.1 - 1 264 Chr. 11 - 62707194 - 62709563 [-] on Build GRCh38
          GenBankmRNA DB108106.1 - 1 213 Chr. 11 - 62707106 - 62709514 [-] on Build GRCh38
          GenBankmRNA DB257059.1 - 1 57 Chr. 11 - 62705473 - 62709358 [-] on Build GRCh38
          GenBankmRNA DA014356.1 - 1 218 Chr. 11 - 62707146 - 62709519 [-] on Build GRCh38
          GenBankmRNA DA379479.1 - 1 236 Chr. 11 - 62707175 - 62709537 [-] on Build GRCh38
          GenBankmRNA DB027858.1 - 1 211 Chr. 11 - 62707108 - 62709511 [-] on Build GRCh38
          GenBankmRNA BX400391.2 - 1 202 Chr. 11 - 62705300 - 62709503 [-] on Build GRCh38
          GenBankmRNA DA381170.1 - 1 209 Chr. 11 - 62707148 - 62709510 [-] on Build GRCh38
          GenBankmRNA BE798840.1 - 1 223 Chr. 11 - 62707123 - 62709503 [-] on Build GRCh38
          GenBankmRNA DB126166.1 - 1 235 Chr. 11 - 62707159 - 62709536 [-] on Build GRCh38
          GenBankmRNA DB127018.1 - 1 209 Chr. 11 - 62707121 - 62709510 [-] on Build GRCh38
          GenBankmRNA DB029908.1 - 1 251 Chr. 11 - 62707143 - 62709551 [-] on Build GRCh38
          GenBankmRNA DB114245.1 - 1 222 Chr. 11 - 62707141 - 62709523 [-] on Build GRCh38
          GenBankmRNA DB184913.1 - 1 223 Chr. 11 - 62707127 - 62709522 [-] on Build GRCh38
          GenBankmRNA BP329574.1 - 1 218 Chr. 11 - 62707115 - 62709519 [-] on Build GRCh38
          GenBankmRNA DB056907.1 - 1 235 Chr. 11 - 62707180 - 62709536 [-] on Build GRCh38
          GenBankmRNA DB193260.1 - 1 110 Chr. 11 - 62705528 - 62709411 [-] on Build GRCh38
          GenBankmRNA CD634220.1 - 1 185 Chr. 11 - 62707208 - 62709486 [-] on Build GRCh38
          GenBankmRNA DB057713.1 - 1 273 Chr. 11 - 62707211 - 62709574 [-] on Build GRCh38
          GenBankmRNA DB058659.1 - 1 264 Chr. 11 - 62707206 - 62709563 [-] on Build GRCh38
          GenBankmRNA CN402425.1 - 1 214 Chr. 11 - 62707193 - 62709513 [-] on Build GRCh38
          GenBankmRNA DA840610.1 - 1 213 Chr. 11 - 62707151 - 62709513 [-] on Build GRCh38
          GenBankmRNA DA906775.1 - 1 147 Chr. 11 - 62705566 - 62709448 [-] on Build GRCh38
          GenBankmRNA DA562868.1 - 1 191 Chr. 11 - 62707149 - 62709492 [-] on Build GRCh38
          GenBankmRNA DA933285.1 - 1 235 Chr. 11 - 62707150 - 62709536 [-] on Build GRCh38
          GenBankmRNA BI857393.1 - 1 203 Chr. 11 - 62705311 - 62709503 [-] on Build GRCh38
          GenBankmRNA AW403174.1 - 1 182 Chr. 11 - 62707233 - 62709476 [-] on Build GRCh38
          GenBankmRNA DA925808.1 - 1 211 Chr. 11 - 62707106 - 62709511 [-] on Build GRCh38
          GenBankmRNA DA582912.1 - 1 223 Chr. 11 - 62707106 - 62709522 [-] on Build GRCh38
          GenBankmRNA BI600943.1 - 1 226 Chr. 11 - 62705403 - 62709526 [-] on Build GRCh38
          GenBankmRNA BP276935.1 - 1 236 Chr. 11 - 62707157 - 62709537 [-] on Build GRCh38
          GenBankmRNA DA924938.1 - 1 240 Chr. 11 - 62707141 - 62709537 [-] on Build GRCh38
          GenBankmRNA DB047290.1 - 1 273 Chr. 11 - 62707196 - 62709574 [-] on Build GRCh38
          GenBankmRNA CD557861.1 - 1 238 Chr. 11 - 62705446 - 62709539 [-] on Build GRCh38
          GenBankmRNA DA523499.1 - 1 235 Chr. 11 - 62707257 - 62709536 [-] on Build GRCh38
          GenBankmRNA BG818222.1 - 1 190 Chr. 11 - 62705310 - 62709476 [-] on Build GRCh38
          GenBankmRNA DB059483.1 - 1 250 Chr. 11 - 62707196 - 62709551 [-] on Build GRCh38
          GenBankmRNA BM454500.1 - 1 184 Chr. 11 - 62702508 - 62709485 [-] on Build GRCh38
          GenBankmRNA BP203300.1 - 1 235 Chr. 11 - 62707200 - 62709536 [-] on Build GRCh38
          GenBankmRNA DA047943.1 - 1 215 Chr. 11 - 62705594 - 62709512 [-] on Build GRCh38
          GenBankmRNA DB271372.1 - 1 231 Chr. 11 - 62707187 - 62709532 [-] on Build GRCh38
          GenBankmRNA DB028798.1 - 1 262 Chr. 11 - 62707147 - 62709563 [-] on Build GRCh38
          GenBankmRNA DA624520.1 - 1 158 Chr. 11 - 62707106 - 62709459 [-] on Build GRCh38
          GenBankmRNA DA588041.1 - 1 222 Chr. 11 - 62707118 - 62709523 [-] on Build GRCh38
          GenBankmRNA DA421825.1 - 1 232 Chr. 11 - 62707168 - 62709532 [-] on Build GRCh38
          GenBankmRNA DA559191.1 - 1 223 Chr. 11 - 62707143 - 62709524 [-] on Build GRCh38
          GenBankmRNA BP345916.1 - 1 73 Chr. 11 - 62705481 - 62709374 [-] on Build GRCh38
          GenBankmRNA DA348487.1 - 1 231 Chr. 11 - 62707132 - 62709532 [-] on Build GRCh38
          GenBankmRNA DA546318.1 - 1 231 Chr. 11 - 62707140 - 62709532 [-] on Build GRCh38
          GenBankmRNA DB267986.1 - 1 218 Chr. 11 - 62707169 - 62709519 [-] on Build GRCh38
          GenBankmRNA DA373651.1 - 1 236 Chr. 11 - 62707166 - 62709537 [-] on Build GRCh38
          GenBankmRNA HY009782.1 - 1 232 Chr. 11 - 62707207 - 62709532 [-] on Build GRCh38
          GenBankmRNA DA622392.1 - 1 220 Chr. 11 - 62707156 - 62709519 [-] on Build GRCh38
          GenBankmRNA BP212607.1 - 1 235 Chr. 11 - 62707140 - 62709537 [-] on Build GRCh38
          GenBankmRNA DA377756.1 - 1 207 Chr. 11 - 62705558 - 62709508 [-] on Build GRCh38
          GenBankmRNA DB026445.1 - 1 235 Chr. 11 - 62707158 - 62709536 [-] on Build GRCh38
          GenBankmRNA BI835156.1 - 1 217 Chr. 11 - 62705368 - 62709518 [-] on Build GRCh38
          GenBankmRNA DB104348.1 - 1 233 Chr. 11 - 62707162 - 62709536 [-] on Build GRCh38
          GenBankmRNA BM926492.1 - 1 48 Chr. 11 - 62692709 - 62709345 [-] on Build GRCh38
          GenBankmRNA BJ993955.1 - 1 222 Chr. 11 - 62707226 - 62709523 [-] on Build GRCh38
          GenBankmRNA DB040395.1 - 1 264 Chr. 11 - 62707199 - 62709564 [-] on Build GRCh38
          GenBankmRNA BI907765.1 - 1 191 Chr. 11 - 62705473 - 62709493 [-] on Build GRCh38
          GenBankmRNA DB053398.1 - 1 231 Chr. 11 - 62707138 - 62709532 [-] on Build GRCh38
          GenBankmRNA DA380099.1 - 1 202 Chr. 11 - 62707139 - 62709503 [-] on Build GRCh38
          GenBankmRNA DA987777.1 - 1 210 Chr. 11 - 62707117 - 62709511 [-] on Build GRCh38
          GenBankmRNA DB248270.1 - 1 236 Chr. 11 - 62705591 - 62709537 [-] on Build GRCh38
          GenBankmRNA DB005105.1 - 1 202 Chr. 11 - 62707144 - 62709503 [-] on Build GRCh38
          GenBankmRNA DA838361.1 - 1 236 Chr. 11 - 62707153 - 62709536 [-] on Build GRCh38
          GenBankmRNA DA377151.1 - 1 235 Chr. 11 - 62707168 - 62709536 [-] on Build GRCh38
          GenBankmRNA BM561833.1 - 285 Chr. - - [-] on Build
          GenBankmRNA DA603204.1 - 1 218 Chr. 11 - 62707147 - 62709519 [-] on Build GRCh38
          GenBankmRNA DA422743.1 - 1 237 Chr. 11 - 62707159 - 62709536 [-] on Build GRCh38
          GenBankmRNA DB002908.1 - 1 235 Chr. 11 - 62707167 - 62709536 [-] on Build GRCh38
          GenBankmRNA CF142050.1 - 1 171 Chr. 11 - 62707133 - 62709464 [-] on Build GRCh38
          GenBankmRNA DA957325.1 - 1 211 Chr. 11 - 62707120 - 62709511 [-] on Build GRCh38
          GenBankmRNA DB497141.1 - 1 214 Chr. 11 - 62707223 - 62709515 [-] on Build GRCh38
          GenBankmRNA CB996968.1 - 1 245 Chr. 11 - 62705409 - 62709523 [-] on Build GRCh38
          GenBankmRNA DB032522.1 - 1 236 Chr. 11 - 62707149 - 62709536 [-] on Build GRCh38
          GenBankmRNA HY167690.1 - 1 211 Chr. 11 - 62707227 - 62709511 [-] on Build GRCh38
          GenBankmRNA BX378643.2 - 1 227 Chr. 11 - 62692696 - 62709528 [-] on Build GRCh38
          GenBankmRNA DA471900.1 - 1 235 Chr. 11 - 62707148 - 62709536 [-] on Build GRCh38
          GenBankmRNA DB192527.1 - 1 205 Chr. 11 - 62707128 - 62709505 [-] on Build GRCh38
          GenBankmRNA DA621561.1 - 1 220 Chr. 11 - 62707149 - 62709521 [-] on Build GRCh38
          GenBankmRNA BU956667.1 - 1 165 Chr. 11 - 62705343 - 62709466 [-] on Build GRCh38
          GenBankmRNA DA558862.1 - 1 236 Chr. 11 - 62707156 - 62709537 [-] on Build GRCh38
          GenBankmRNA DA746943.1 - 1 219 Chr. 11 - 62707131 - 62709519 [-] on Build GRCh38
          GenBankmRNA HY073949.1 - 1 232 Chr. 11 - 62707202 - 62709532 [-] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          lipid catabolic process
          lipid storage
          lipid particle organization
          fat cell differentiation
          negative regulation of lipid catabolic process
          molecular_function
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0