Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NOG SIRNA Assays ›
Gene Symbol:
NOG
Gene Name:
noggin
Gene Aliases:
SYM1, SYNS1, SYNS1A
Chromosome Location:
Chr.17: 56593699 - 56595590 on Build GRCh38
UniGene ID:
Hs.248201
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s534107
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NOG
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005450.4
NP_005441.1
1
1481
Back To Top
More Information
Panther Classification:
Molecular Function -
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
skeletal system development
osteoblast differentiation
in utero embryonic development
endoderm formation
mesoderm formation
epithelial to mesenchymal transition
neural plate morphogenesis
neural tube closure
nervous system development
motor neuron axon guidance
dorsal/ventral pattern formation
spinal cord development
cell differentiation in hindbrain
pituitary gland development
negative regulation of cell migration
BMP signaling pathway
negative regulation of BMP signaling pathway
somatic stem cell population maintenance
wound healing
middle ear morphogenesis
embryonic digit morphogenesis
regulation of neuron differentiation
negative regulation of osteoblast differentiation
positive regulation of transcription from RNA polymerase II promoter
axial mesoderm development
notochord morphogenesis
embryonic skeletal system development
negative regulation of astrocyte differentiation
positive regulation of epithelial cell proliferation
cartilage development
negative regulation of cardiac muscle cell proliferation
limb development
embryonic skeletal joint morphogenesis
negative regulation of cytokine activity
face morphogenesis
negative regulation of pathway-restricted SMAD protein phosphorylation
lung morphogenesis
prostatic bud formation
ureteric bud formation
fibroblast growth factor receptor signaling pathway involved in neural plate anterior/posterior pattern formation
negative regulation of cartilage development
somite development
negative regulation of canonical Wnt signaling pathway
positive regulation of branching involved in ureteric bud morphogenesis
positive regulation of glomerulus development
regulation of fibroblast growth factor receptor signaling pathway involved in neural plate anterior/posterior pattern formation
negative regulation of apoptotic signaling pathway
protein binding
cytokine binding
protein complex binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0