Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NF1 SIRNA Assays ›
          Gene Symbol:
          NF1
          Gene Name:
          neurofibromin 1
          Gene Aliases:
          NFNS, VRNF, WSS
          Chromosome Location:
          Chr.17: 31094927 - 31377677 on Build GRCh38
          UniGene ID:
          Hs.113577
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s534236

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000267.3 NP_000258.1 2 508
          NM_001042492.2 NP_001035957.1 2 508
          NM_001128147.2 NP_001121619.1 2 508
          XM_005257983.2 XP_005258040.1 2 458
          XM_005257984.2 XP_005258041.1 2 458
          XM_006721922.2 XP_006721985.1 2 458
          XM_006721924.2 XP_006721987.1 2 458
          XM_006721925.2 XP_006721988.1 2 458
          XM_006721926.3 XP_006721989.1 2 458
          XM_011524852.2 XP_011523154.1 2 458
          XM_011524857.2 XP_011523159.1 2 458
          XM_017024689.1 XP_016880178.1 2 458
          XM_017024690.1 XP_016880179.1 2 458
          GenBank mRNA AK289936.1 - 2 458
          BC144643.1 - 2 227
          D12625.1 - 2 508
          D42072.1 - 2 190
          DA299151.1 - 2 458
          DB094344.1 - 2 458
          M82814.1 - 2 336
          M89914.1 - 2 322

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          osteoblast differentiation
          metanephros development
          response to hypoxia
          liver development
          negative regulation of endothelial cell proliferation
          positive regulation of endothelial cell proliferation
          regulation of cell-matrix adhesion
          negative regulation of cell-matrix adhesion
          negative regulation of protein kinase activity
          cell communication
          Ras protein signal transduction
          negative regulation of neuroblast proliferation
          brain development
          peripheral nervous system development
          heart development
          skeletal muscle tissue development
          visual learning
          extrinsic apoptotic signaling pathway via death domain receptors
          regulation of gene expression
          Schwann cell development
          phosphatidylinositol 3-kinase signaling
          negative regulation of angiogenesis
          spinal cord development
          amygdala development
          forebrain astrocyte development
          neural tube development
          cerebral cortex development
          myelination in peripheral nervous system
          actin cytoskeleton organization
          extracellular matrix organization
          collagen fibril organization
          adrenal gland development
          negative regulation of cell migration
          regulation of synaptic transmission, GABAergic
          cellular response to heat
          negative regulation of Rac protein signal transduction
          wound healing
          negative regulation of transcription factor import into nucleus
          positive regulation of apoptotic process
          regulation of GTPase activity
          negative regulation of MAP kinase activity
          negative regulation of MAPK cascade
          pigmentation
          positive regulation of neuron apoptotic process
          regulation of blood vessel endothelial cell migration
          positive regulation of GTPase activity
          regulation of bone resorption
          negative regulation of osteoclast differentiation
          regulation of glial cell differentiation
          positive regulation of adenylate cyclase activity
          regulation of angiogenesis
          negative regulation of Ras protein signal transduction
          negative regulation of neurotransmitter secretion
          negative regulation of fibroblast proliferation
          regulation of long-term neuronal synaptic plasticity
          sympathetic nervous system development
          camera-type eye morphogenesis
          negative regulation of astrocyte differentiation
          negative regulation of oligodendrocyte differentiation
          smooth muscle tissue development
          artery morphogenesis
          forebrain morphogenesis
          cognition
          gamma-aminobutyric acid secretion, neurotransmission
          glutamate secretion, neurotransmission
          observational learning
          regulation of long-term synaptic potentiation
          positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          GTPase activator activity
          protein binding
          phosphatidylethanolamine binding
          phosphatidylcholine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0