Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RSPH9 SIRNA Assays ›
          Gene Symbol:
          RSPH9
          Gene Name:
          radial spoke head 9 homolog
          Gene Aliases:
          C6orf206, CILD12, MRPS18AL1
          Chromosome Location:
          Chr.6: 43645030 - 43671011 on Build GRCh38
          UniGene ID:
          Hs.534585
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s534377

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RSPH9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001193341.1 NP_001180270.1 1 166
          NM_152732.4 NP_689945.2 1 166
          XM_005248901.3 XP_005248958.1 1 139
          XM_006715014.1 XP_006715077.1 1 166
          XM_017010426.1 XP_016865915.1 1 139
          GenBank mRNA AK055407.1 - 1 106
          AK291399.1 - 1 139
          BC029519.1 - 1 123 5296237
          DA486918.1 - 1 167
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CV024680.1 - 1 97 Chr. 6 - 43645099 - 43656665 [+] on Build GRCh38
          GenBankmRNA BP373299.1 - 1 152 Chr. 6 - 43645044 - 43655692 [+] on Build GRCh38
          GenBankmRNA BU196545.1 - 1 146 Chr. 6 - 43645050 - 43670997 [+] on Build GRCh38
          GenBankmRNA AA417188.1 - 1 23 Chr. 6 - 43645173 - 43655623 [+] on Build GRCh38
          GenBankmRNA BX357148.2 - 1 98 Chr. 6 - 43645098 - 43671005 [+] on Build GRCh38
          GenBankmRNA D61437.1 - 1 86 Chr. 6 - 43645110 - 43650518 [+] on Build GRCh38
          GenBankmRNA BU171360.1 - 1 125 Chr. 6 - 43645073 - 43656705 [+] on Build GRCh38
          GenBankmRNA R12615.1 - 148 Chr. - - [-] on Build
          GenBankmRNA DB501550.1 - 1 123 Chr. 6 - 43645073 - 43655653 [+] on Build GRCh38
          GenBankmRNA T78771.1 - 1 128 Chr. 6 - 43645072 - 43650421 [+] on Build GRCh38
          GenBankmRNA CD694011.1 - 1 167 Chr. 6 - 43645070 - 43650513 [+] on Build GRCh38
          GenBankmRNA DA803796.1 - 1 123 Chr. 6 - 43645073 - 43651889 [+] on Build GRCh38
          GenBankmRNA DA514830.1 - 1 106 Chr. 6 - 43645090 - 43656639 [+] on Build GRCh38
          GenBankmRNA HY040250.1 - 1 123 Chr. 6 - 43645073 - 43656611 [+] on Build GRCh38
          GenBankmRNA BQ680488.1 - 1 171 Chr. 6 - 43645047 - 43671008 [+] on Build GRCh38
          GenBankmRNA DA791521.1 - 1 139 Chr. 6 - 43645057 - 43655624 [+] on Build GRCh38
          GenBankmRNA BI826340.1 - 1 112 Chr. 6 - 43645083 - 43670927 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cilium movement
          axoneme assembly
          motile cilium assembly
          cilium movement involved in cell motility
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0