Search
Search

POMK| Gene Symbol: POMK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277971.1 | NP_001264900.1 | 4 | 492 | ||
| NM_032237.4 | NP_115613.1 | 5 | 588 | |||
| GenBank mRNA | BC101548.1 | - | 3 | 359 | 8069054 | |
| BC113703.1 | - | 3 | 359 | 8322755 | ||
| DA383787.1 | - | 5 | 580 | |||
| DA818523.1 | - | 4 | 483 | |||