Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CD36 SIRNA Assays ›
          Gene Symbol:
          CD36
          Gene Name:
          CD36 molecule
          Gene Aliases:
          BDPLT10, CHDS7, FAT, GP3B, GP4, GPIV, PASIV, SCARB3
          Chromosome Location:
          Chr.7: 80602188 - 80679277 on Build GRCh38
          UniGene ID:
          Hs.120949
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s534751

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CD36
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000072.3 NP_000063.2 4, 5 698
          NM_001001547.2 NP_001001547.1 4, 5 659
          NM_001001548.2 NP_001001548.1 4, 5 698
          NM_001127443.1 NP_001120915.1 3, 4 404
          NM_001127444.1 NP_001120916.1 2, 3 579
          NM_001289908.1 NP_001276837.1 2, 3 372
          NM_001289909.1 NP_001276838.1 3, 4 463
          NM_001289911.1 NP_001276840.1 2, 3 337
          NR_110501.1 3, 4 463
          XM_005250713.1 XP_005250770.1 4, 5 640
          XM_005250714.1 XP_005250771.1 4, 5 607
          XM_005250715.4 XP_005250772.1 4, 5 765
          GenBank mRNA AK096858.1 - 7, 8 968
          AK129899.1 - 4, 5 640
          AK300101.1 - 2, 3 337
          AK312920.1 - 4, 5 622
          BC008406.1 - 4, 5 573 4244251
          DA566475.1 - 3, 4 404
          HM217020.1 - 2, 3 381
          HM217021.1 - 2, 3 381
          HM217022.1 - 2, 3 381
          HM217023.1 - 2, 3 381
          HM217024.1 - 2, 3 381
          HM217025.1 - 2, 3 381
          HM217026.1 - 2, 3 372
          KU177961.1 - 2, 3 284
          L06850.1 - 3, 4 416
          M24795.1 - 4, 5 494
          M98398.1 - 4, 5 538
          M98399.1 - 4, 5 538
          S67532.1 - 3, 4 420

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          positive regulation of cell-matrix adhesion
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          platelet degranulation
          MyD88-dependent toll-like receptor signaling pathway
          lipid metabolic process
          receptor-mediated endocytosis
          phagocytosis, recognition
          immune response
          cell adhesion
          cell surface receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          nitric oxide mediated signal transduction
          blood coagulation
          positive regulation of macrophage derived foam cell differentiation
          positive regulation of cholesterol storage
          lipid storage
          cGMP-mediated signaling
          positive regulation of blood coagulation
          intestinal cholesterol absorption
          cholesterol transport
          receptor internalization
          positive regulation of interleukin-12 production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          response to lipid
          triglyceride transport
          plasma lipoprotein particle clearance
          low-density lipoprotein particle clearance
          response to stilbenoid
          toll-like receptor TLR6:TLR2 signaling pathway
          lipoprotein transport
          negative regulation of transcription factor import into nucleus
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          apoptotic cell clearance
          negative regulation of growth of symbiont in host
          long-chain fatty acid import
          interleukin-1 beta secretion
          positive regulation of peptidyl-tyrosine phosphorylation
          defense response to Gram-positive bacterium
          intestinal absorption
          sensory perception of taste
          low-density lipoprotein particle mediated signaling
          positive regulation of phagocytosis, engulfment
          positive regulation of macrophage cytokine production
          positive regulation of ERK1 and ERK2 cascade
          cholesterol import
          response to fatty acid
          response to linoleic acid
          cellular response to lipopolysaccharide
          cellular response to lipoteichoic acid
          cellular response to low-density lipoprotein particle stimulus
          cellular response to hydroperoxide
          cellular response to diacyl bacterial lipopeptide
          positive regulation of NLRP3 inflammasome complex assembly
          amyloid fibril formation
          regulation of removal of superoxide radicals
          positive regulation of blood microparticle formation
          positive regulation of reactive oxygen species metabolic process
          regulation of energy homeostasis
          low-density lipoprotein receptor activity
          protein binding
          high-density lipoprotein particle binding
          lipid binding
          low-density lipoprotein particle binding
          transforming growth factor beta binding
          thrombospondin receptor activity
          lipoteichoic acid receptor activity
          lipoprotein particle binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0