Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LHB SIRNA Assays ›
          Gene Symbol:
          LHB CGB1 CGB2 CGB3 CGB5 CGB7 CGB8
          Gene Name:
          luteinizing hormone beta polypeptide
          chorionic gonadotropin beta subunit 1
          chorionic gonadotropin beta subunit 2
          chorionic gonadotropin beta subunit 3
          chorionic gonadotropin beta subunit 5
          chorionic gonadotropin beta subunit 7
          chorionic gonadotropin beta subunit 8
          Gene Aliases:
          CGB4, HH23, LSH-B, LSH-beta
          -
          -
          CGB, CGB5, CGB7, CGB8, hCGB
          CGB, HCG, hCGB
          CG-beta-a, CGB6
          -
          Chromosome Location:
          Chr.19: 49015980 - 49017090 on Build GRCh38
          Chromosome Location:
          Chr.19: 49035569 - 49036934 on Build GRCh38
          Chromosome Location:
          Chr.19: 49031873 - 49033238 on Build GRCh38
          Chromosome Location:
          Chr.19: 49022869 - 49024375 on Build GRCh38
          Chromosome Location:
          Chr.19: 49043845 - 49045311 on Build GRCh38
          Chromosome Location:
          Chr.19: 49054274 - 49058346 on Build GRCh38
          Chromosome Location:
          Chr.19: 49047638 - 49049111 on Build GRCh38
          UniGene ID:
          Hs.154704
          Hs.659014
          Hs.567650
          Hs.446683
          Hs.172944
          Hs.681647
          Hs.659014
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s535191

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LHB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000894.2 NP_000885.1 2 161
          XM_011526975.1 XP_011525277.1 1 235
          Gene Symbol: CGB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033377.1 NP_203695.2 2 369
          XM_017026233.1 XP_016881722.1 1 211
          GenBank mRNA BC107144.1 - 2 507
          BC126460.1 - 2 251
          Gene Symbol: CGB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001319065.1 NP_001305994.1 2 496
          NM_033378.1 NP_203696.2 2 369
          XM_005258479.3 XP_005258536.1 1 302
          GenBank mRNA BC069367.1 - 2 264 7262156
          BC107143.1 - 2 496
          Gene Symbol: CGB3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000737.3 NP_000728.1 2 556
          GenBank mRNA AK291552.1 - 2 514
          BC022796.1 - 2 494 4811294
          BC069526.1 - 2 161 7262120
          BC106059.1 - 2 502 4992804
          BX334905.2 - 2 561
          J00117.1 - 2 177
          Gene Symbol: CGB5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033043.1 NP_149032.1 2 517
          GenBank mRNA BC006290.2 - 2 478 3960125
          BC051378.1 - 2 478 3956670
          BC103970.1 - 2 248 40014677
          BC103971.1 - 2 244 40014679
          BC106723.1 - 2 162 40014469
          BC106724.1 - 2 161 40014470
          BT006890.1 - 1 152
          Gene Symbol: CGB7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033142.1 NP_149133.1 2 517
          XM_017027502.1 XP_016882991.1 5 1864
          GenBank mRNA AK092722.1 - 4 1587
          AK125108.1 - 4 3024
          Gene Symbol: CGB8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033183.2 NP_149439.1 2 524
          GenBank mRNA BC030994.1 - 2 519 4605021
          BC103969.1 - 2 244 40014676
          BC128603.1 - 2 244 40002893
          BX118262.1 - 2 524
          BX380841.2 - 2 504

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          progesterone biosynthetic process
          signal transduction
          cell-cell signaling
          male gonad development
          peptide hormone processing
          receptor binding
          hormone activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0