Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CGB8 SIRNA Assays ›
          Gene Symbol:
          CGB8 CGB1 CGB2 CGB3 CGB5 CGB7 LHB
          Gene Name:
          chorionic gonadotropin beta subunit 8
          chorionic gonadotropin beta subunit 1
          chorionic gonadotropin beta subunit 2
          chorionic gonadotropin beta subunit 3
          chorionic gonadotropin beta subunit 5
          chorionic gonadotropin beta subunit 7
          luteinizing hormone beta polypeptide
          Gene Aliases:
          -
          -
          -
          CGB, CGB5, CGB7, CGB8, hCGB
          CGB, HCG, hCGB
          CG-beta-a, CGB6
          CGB4, HH23, LSH-B, LSH-beta
          Chromosome Location:
          Chr.19: 49047638 - 49049111 on Build GRCh38
          Chromosome Location:
          Chr.19: 49035569 - 49036934 on Build GRCh38
          Chromosome Location:
          Chr.19: 49031873 - 49033238 on Build GRCh38
          Chromosome Location:
          Chr.19: 49022869 - 49024375 on Build GRCh38
          Chromosome Location:
          Chr.19: 49043845 - 49045311 on Build GRCh38
          Chromosome Location:
          Chr.19: 49054274 - 49058346 on Build GRCh38
          Chromosome Location:
          Chr.19: 49015980 - 49017090 on Build GRCh38
          UniGene ID:
          Hs.659014
          Hs.659014
          Hs.567650
          Hs.446683
          Hs.172944
          Hs.681647
          Hs.154704
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s535194

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CGB8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033183.2 NP_149439.1 3 873
          GenBank mRNA BC030994.1 - 3 868 4605021
          BC103969.1 - 3 593 40014676
          BC128603.1 - 3 593 40002893
          Gene Symbol: CGB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033377.1 NP_203695.2 3 718
          XM_017026233.1 XP_016881722.1 2 560
          Gene Symbol: CGB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001319065.1 NP_001305994.1 3 845
          NM_033378.1 NP_203696.2 3 718
          XM_005258479.3 XP_005258536.1 2 651
          Gene Symbol: CGB3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000737.3 NP_000728.1 3 905
          GenBank mRNA AK291552.1 - 3 863
          BC022796.1 - 3 843 4811294
          BC069526.1 - 3 510 7262120
          BC106059.1 - 3 851 4992804
          BX334905.2 - 3 909
          J00117.1 - 3 526
          Gene Symbol: CGB5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033043.1 NP_149032.1 3 866
          GenBank mRNA BC006290.2 - 3 827 3960125
          BC051378.1 - 3 827 3956670
          BC103970.1 - 3 597 40014677
          BC103971.1 - 3 593 40014679
          Gene Symbol: CGB7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033142.1 NP_149133.1 3 866
          XM_017027502.1 XP_016882991.1 6 2213
          GenBank mRNA AK092722.1 - 5 1936
          AK125108.1 - 5 3373
          Gene Symbol: LHB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000894.2 NP_000885.1 3 509
          XM_011526975.1 XP_011525277.1 2 583

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          progesterone biosynthetic process
          signal transduction
          cell-cell signaling
          male gonad development
          peptide hormone processing
          hormone activity
          receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0