Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL6ST SIRNA Assays ›
          Gene Symbol:
          IL6ST
          Gene Name:
          interleukin 6 signal transducer
          Gene Aliases:
          CD130, CDW130, GP130, IL-6RB
          Chromosome Location:
          Chr.5: 55935095 - 55994993 on Build GRCh38
          UniGene ID:
          Hs.532082
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s535300

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IL6ST
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001190981.1 NP_001177910.1 7 1070
          NM_002184.3 NP_002175.2 7 1070
          NM_175767.2 NP_786943.1 7 1070
          NR_120480.1 7 1070
          XM_017009441.1 XP_016864930.1 6 926
          XM_017009442.1 XP_016864931.1 6 949
          GenBank mRNA AB015706.1 - 6 849
          AB102801.1 - 3 259
          AB102802.1 - 5 757
          BC117402.1 - 6 869 40125953
          BC117404.1 - 6 869 40125955
          HQ628629.1 - 5 757
          HQ628630.1 - 5 757
          M57230.1 - 7 1012

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of acute inflammatory response
          positive regulation of adaptive immune response
          glycogen metabolic process
          positive regulation of cell proliferation
          regulation of Notch signaling pathway
          positive regulation of vascular endothelial growth factor production
          positive regulation of cardiac muscle hypertrophy
          viral process
          cytokine-mediated signaling pathway
          response to cytokine
          interleukin-11-mediated signaling pathway
          oncostatin-M-mediated signaling pathway
          positive regulation of T cell proliferation
          positive regulation of tyrosine phosphorylation of Stat1 protein
          positive regulation of tyrosine phosphorylation of Stat3 protein
          negative regulation of apoptotic process
          positive regulation of osteoblast differentiation
          positive regulation of astrocyte differentiation
          leukemia inhibitory factor signaling pathway
          interleukin-6-mediated signaling pathway
          interleukin-27-mediated signaling pathway
          ciliary neurotrophic factor-mediated signaling pathway
          ciliary neurotrophic factor receptor activity
          interleukin-6 receptor activity
          interleukin-11 receptor activity
          leukemia inhibitory factor receptor activity
          oncostatin-M receptor activity
          ciliary neurotrophic factor receptor binding
          interleukin-6 receptor binding
          protein binding
          growth factor binding
          interleukin-11 binding
          interleukin-6 binding
          protein homodimerization activity
          interleukin-27 receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0