Search
Search

| Gene Symbol: OR5H15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005515.1 | NP_001005515.1 | 1 | 517 | ||
| Gene Symbol: OR5H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005338.1 | NP_001005338.1 | 1 | 517 | ||
| GenBank mRNA | X64988.1 | - | 1 | 143 | ||