Search
Search

FAM45A| Gene Symbol: FAM45A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303111.1 | NP_001290040.1 | 10 | 2309 | ||
| NM_001303112.1 | NP_001290041.1 | 9 | 2056 | |||
| NM_001303113.1 | NP_001290042.1 | 8 | 1975 | |||
| NM_207009.3 | NP_996892.1 | 9 | 2104 | |||
| NR_130122.1 | 8 | 1992 | ||||
| XM_006717849.3 | XP_006717912.1 | 9 | 2124 | |||
| XM_006717850.3 | XP_006717913.1 | 10 | 2366 | |||
| XM_006717851.3 | XP_006717914.1 | 10 | 2346 | |||
| XM_006717852.3 | XP_006717915.1 | 10 | 2330 | |||
| XM_017016261.1 | XP_016871750.1 | 10 | 2839 | |||
| XM_017016262.1 | XP_016871751.1 | 11 | 2554 | |||
| XM_017016263.1 | XP_016871752.1 | 11 | 2535 | |||
| XM_017016264.1 | XP_016871753.1 | 9 | 1499 | |||
| XM_017016265.1 | XP_016871754.1 | 10 | 2350 | |||
| XM_017016266.1 | XP_016871755.1 | 11 | 2531 | |||
| XM_017016267.1 | XP_016871756.1 | 11 | 2515 | |||
| GenBank mRNA | AK027029.1 | - | 7 | 2204 | ||