Search
Search

GALNTL6| Gene Symbol: GALNTL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034845.2 | NP_001030017.2 | 4 | 1082 | ||
| XM_011531993.2 | XP_011530295.1 | 3 | 420 | |||
| XM_017008243.1 | XP_016863732.1 | 4 | 1816 | |||
| XM_017008244.1 | XP_016863733.1 | 3 | 1188 | |||
| GenBank mRNA | AJ626725.1 | - | 3 | 295 | ||
| AK295464.1 | - | 3 | 499 | |||
| BC047551.1 | - | 4 | 1003 | 5301536 | ||
| GU220060.1 | - | 3 | 346 | |||