Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other POLR2J3 SIRNA Assays ›
          Gene Symbol:
          POLR2J3 POLR2J POLR2J2 POLR2J4
          Gene Name:
          RNA polymerase II subunit J3
          RNA polymerase II subunit J
          RNA polymerase II subunit J2
          RNA polymerase II subunit J4, pseudogene
          Gene Aliases:
          POLR2J2, RPB11b1, RPB11b2
          POLR2J1, RPB11, RPB11A, RPB11m, hRPB14
          HRPB11B, RPB11b1
          RPB11-phi
          Chromosome Location:
          Chr.7: 102537919 - 102572660 on Build GRCh38
          Chromosome Location:
          Chr.7: 102473100 - 102478999 on Build GRCh38
          Chromosome Location:
          Chr.7: 102636748 - 102671729 on Build GRCh38
          Chromosome Location:
          Chr.7: 43940895 - 44019194 on Build GRCh38
          UniGene ID:
          Hs.610255
          Hs.654952
          Hs.610255
          Hs.657028
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID s54728

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: POLR2J3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001097615.2 NP_001091084.2 2 187
          XM_017012372.1 XP_016867861.1 2 226
          XM_017012373.1 XP_016867862.1 2 226
          GenBank mRNA AF468111.1 - 2 133
          AJ277740.1 - 2 87
          AJ277741.1 - 2 87
          AK309487.1 - 2 149
          BC141857.1 - 2 146
          DA574442.1 - 2 150
          DA580738.1 - 2 149
          DB106772.1 - 2 188
          Gene Symbol: POLR2J
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006234.4 NP_006225.1 2 164
          XM_005250452.4 XP_005250509.1 2 229
          XM_017012353.1 XP_016867842.1 2 161
          GenBank mRNA AK290515.1 - 2 161
          BC024165.1 - 2 130 4069069
          BC040116.1 - 2 125
          BC065711.1 - 2 142 4133200
          BC139902.1 - 2 127 40147225
          BC141830.1 - 2 126 40148671
          BM545237.1 - 2 282
          L37127.1 - 2 151
          X82385.1 - 2 151
          X98433.1 - 2 118
          Gene Symbol: POLR2J2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AB209398.1 - 2 153
          AJ277739.1 - 2 87
          BC056864.1 - 2 105
          Gene Symbol: POLR2J4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003655.3 2 171
          GenBank mRNA AK296066.1 - 2 152
          BC017250.2 - 2 147
          BC017341.2 - 2 135
          BC062722.1 - 2 130
          BC105584.1 - 2 153
          BX537878.1 - 2 126
          BX538054.1 - 2 128
          DA147922.1 - 2 171

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-directed RNA polymerase RNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          mRNA splicing, via spliceosome
          transcription-coupled nucleotide-excision repair
          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          transcription elongation from RNA polymerase II promoter
          7-methylguanosine mRNA capping
          fibroblast growth factor receptor signaling pathway
          gene expression
          gene silencing by RNA
          somatic stem cell population maintenance
          snRNA transcription from RNA polymerase II promoter
          positive regulation of viral transcription
          DNA binding
          DNA-directed RNA polymerase activity
          protein dimerization activity
          RNA polymerase II activity
          protein binding
          LRR domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0