Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KMT2C SIRNA Assays ›
Gene Symbol:
KMT2C
LOC107983967
Gene Name:
lysine methyltransferase 2C
histone-lysine N-methyltransferase 2C-like
Gene Aliases:
HALR, MLL3
-
Chromosome Location:
Chr.7: 152134925 - 152436642 on Build GRCh38
Chromosome Location:
Chr.1: 143464779 - 143476935 on Build GRCh38
UniGene ID:
Hs.647120
-
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s55932
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KMT2C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_170606.2
NP_733751.2
18, 19
3196
XM_005250025.4
XP_005250082.1
19, 20
3002
XM_005250026.3
XP_005250083.1
19, 20
3036
XM_005250027.4
XP_005250084.1
18, 19
3210
XM_005250028.4
XP_005250085.1
18, 19
3210
XM_005250031.4
XP_005250088.1
18, 19
3210
XM_006716077.3
XP_006716140.1
18, 19
3210
XM_006716078.3
XP_006716141.1
18, 19
3210
XM_006716079.3
XP_006716142.1
18, 19
3210
XM_011516450.2
XP_011514752.1
18, 19
3210
XM_011516451.2
XP_011514753.1
17, 18
3090
XM_011516452.2
XP_011514754.1
18, 19
3210
XM_011516453.2
XP_011514755.1
18, 19
3210
XM_011516454.2
XP_011514756.1
13, 14
2205
XM_011516456.2
XP_011514758.1
18, 19
3210
XM_017012480.1
XP_016867969.1
19, 20
3039
XM_017012481.1
XP_016867970.1
19, 20
2999
XM_017012482.1
XP_016867971.1
18, 19
3210
XM_017012483.1
XP_016867972.1
18, 19
3210
XM_017012484.1
XP_016867973.1
18, 19
3177
XM_017012485.1
XP_016867974.1
18, 19
3207
XM_017012486.1
XP_016867975.1
18, 19
3210
XM_017012487.1
XP_016867976.1
18, 19
3178
XM_017012488.1
XP_016867977.1
18, 19
3210
GenBank mRNA
AB040939.2
-
7, 8
1312
AF264750.2
-
4, 5
523
AK025911.1
-
8, 9
1422
AK095628.1
-
11, 12
1918
AY024361.1
-
18, 19
3157
BC151837.1
-
7, 8
1312
Gene Symbol: LOC107983967
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_017003062.1
XP_016858551.1
4, 5
399
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008762670.
This siRNA also targets the following transcripts in Mouse: NM_001081383, AY036886, AK087368, AY138582, BC037044.
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
histone H3-K4 methylation
DNA binding
protein binding
zinc ion binding
transferase activity, transferring acyl groups
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K4 specific)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0