Search
Search

| Gene Symbol: GBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002053.2 | NP_002044.2 | 10 | 1788 | ||
| GenBank mRNA | AB208912.1 | - | 7 | 3390 | ||
| AK291783.1 | - | 10 | 1636 | |||
| AK298032.1 | - | 7 | 962 | |||
| AK303893.1 | - | 10 | 1591 | |||
| AY920436.1 | - | 9 | 1572 | |||
| BC002666.2 | - | 10 | 1788 | 3606865 | ||
| BT006847.1 | - | 9 | 1568 | |||
| M55542.1 | - | 10 | 1636 | |||
| Gene Symbol: GBP1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003133.2 | 5 | 654 | |||
| GenBank mRNA | AK309962.1 | - | 5 | 654 | ||